Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 93593591-93593830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR2F1
Entrez Gene ID
7025
Gene Name
nuclear receptor subfamily 2 group F member 1
Gene Aliases
BBOAS, BBSOAS, COUP-TFI, EAR-3, EAR3, ERBAL3, NR2F2, SVP44, TCFCOUP1, TFCOUP1
UniGene
Hs.519445
Gene Chromosome Location
Chr.5: 93583337-93594613 on Build GRCh38
Transcripts
NM_005654.5 XM_017009797.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6729169 c.1050C>T p.C350C Chr.5: 93593620-93593620 on Build GRCh38
6729170 c.1207A>G p.I403V Chr.5: 93593777-93593777 on Build GRCh38
5579121 c.1041G>A p.K347K Chr.5: 93593611-93593611 on Build GRCh38
5684913 c.1030C>A p.L344M Chr.5: 93593600-93593600 on Build GRCh38
1071244 c.1043C>T p.S348L Chr.5: 93593613-93593613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008981901 Chr.5: 93593745-93593745 on Build GRCh38
rs946373674 Chr.5: 93593777-93593777 on Build GRCh38
rs370092446 Chr.5: 93593725-93593725 on Build GRCh38
rs142689077 Chr.5: 93593614-93593614 on Build GRCh38
rs763566932 Chr.5: 93593635-93593635 on Build GRCh38
rs746100563 Chr.5: 93593717-93593717 on Build GRCh38
rs377161852 Chr.5: 93593711-93593711 on Build GRCh38
rs1028527666 Chr.5: 93593649-93593649 on Build GRCh38
rs774849931 Chr.5: 93593779-93593779 on Build GRCh38
rs747029125 Chr.5: 93593710-93593710 on Build GRCh38
rs570838351 Chr.5: 93593702-93593702 on Build GRCh38
rs1015063343 Chr.5: 93593764-93593764 on Build GRCh38
rs781436664 Chr.5: 93593716-93593716 on Build GRCh38
rs530910180 Chr.5: 93593620-93593620 on Build GRCh38
rs755893427 Chr.5: 93593692-93593692 on Build GRCh38
rs750040062 Chr.5: 93593789-93593789 on Build GRCh38
rs373277412 Chr.5: 93593805-93593805 on Build GRCh38
rs767095926 Chr.5: 93593656-93593656 on Build GRCh38
rs756887222 Chr.5: 93593647-93593647 on Build GRCh38
rs779735831 Chr.5: 93593698-93593698 on Build GRCh38
rs144876856 Chr.5: 93593719-93593719 on Build GRCh38
rs751140216 Chr.5: 93593636-93593636 on Build GRCh38
rs1050931871 Chr.5: 93593624-93593624 on Build GRCh38
rs755004144 Chr.5: 93593704-93593704 on Build GRCh38
rs775761102 Chr.5: 93593720-93593720 on Build GRCh38
rs138827038 Chr.5: 93593767-93593767 on Build GRCh38
rs776020140 Chr.5: 93593621-93593621 on Build GRCh38
rs761244180 Chr.5: 93593782-93593782 on Build GRCh38
rs371426115 Chr.5: 93593662-93593662 on Build GRCh38
rs766964375 Chr.5: 93593784-93593784 on Build GRCh38
rs764011372 Chr.5: 93593666-93593666 on Build GRCh38
rs550670145 Chr.5: 93593629-93593629 on Build GRCh38

View Full Product Details