Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 145918593-145918334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX11B
Entrez Gene ID
8799
Gene Name
peroxisomal biogenesis factor 11 beta
Gene Aliases
PEX11-BETA, PEX14B
UniGene
Hs.714608
Gene Chromosome Location
Chr.1: 145911348-145918924 on Build GRCh38
Transcripts
NM_001184795.1 NM_003846.2 NR_073491.1 NR_073492.1 NR_073493.2

View Full Product Details