Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 86657977-86658222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPN4
Entrez Gene ID
94233
Gene Name
opsin 4
Gene Aliases
MOP
UniGene
Hs.283922
Gene Chromosome Location
Chr.10: 86653495-86666460 on Build GRCh38
Transcripts
NM_001030015.2 NM_033282.3 XM_017016955.1 XM_017016956.1 XM_017016957.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5900195 c.365A>G p.N122S Chr.10: 86658073-86658073 on Build GRCh38
4709531 c.379G>A p.D127N Chr.10: 86658087-86658087 on Build GRCh38
3441178 c.324G>A p.Q108Q Chr.10: 86658032-86658032 on Build GRCh38
1349384 c.442C>T p.L148F Chr.10: 86658150-86658150 on Build GRCh38
5837400 c.388A>G p.M130V Chr.10: 86658096-86658096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766640490 Chr.10: 86658031-86658031 on Build GRCh38
rs757119574 Chr.10: 86658199-86658199 on Build GRCh38
rs756013544 Chr.10: 86658066-86658066 on Build GRCh38
rs761987702 Chr.10: 86658087-86658087 on Build GRCh38
rs994273641 Chr.10: 86658086-86658086 on Build GRCh38
rs996086270 Chr.10: 86658029-86658029 on Build GRCh38
rs777454188 Chr.10: 86658033-86658035 on Build GRCh38
rs1041022881 Chr.10: 86658071-86658071 on Build GRCh38
rs760799384 Chr.10: 86658003-86658003 on Build GRCh38
rs751292318 Chr.10: 86658032-86658032 on Build GRCh38
rs776952356 Chr.10: 86658081-86658081 on Build GRCh38
rs751704144 Chr.10: 86658075-86658075 on Build GRCh38
rs375300822 Chr.10: 86658046-86658046 on Build GRCh38
rs767191686 Chr.10: 86658167-86658167 on Build GRCh38
rs758805076 Chr.10: 86658022-86658022 on Build GRCh38
rs777698950 Chr.10: 86658054-86658054 on Build GRCh38
rs747304015 Chr.10: 86658078-86658078 on Build GRCh38
rs749165210 Chr.10: 86658061-86658061 on Build GRCh38
rs773828378 Chr.10: 86658112-86658112 on Build GRCh38
rs751851259 Chr.10: 86658134-86658134 on Build GRCh38
rs776258464 Chr.10: 86658115-86658116 on Build GRCh38
rs773981276 Chr.10: 86658077-86658077 on Build GRCh38
rs765351988 Chr.10: 86658096-86658096 on Build GRCh38
rs754559194 Chr.10: 86658042-86658042 on Build GRCh38
rs368882005 Chr.10: 86658080-86658080 on Build GRCh38
rs753850060 Chr.10: 86658192-86658192 on Build GRCh38
rs890024153 Chr.10: 86658060-86658060 on Build GRCh38
rs571378526 Chr.10: 86658110-86658110 on Build GRCh38
rs527421661 Chr.10: 86658021-86658021 on Build GRCh38
rs199760115 Chr.10: 86658013-86658013 on Build GRCh38
rs1048938714 Chr.10: 86658051-86658051 on Build GRCh38
rs375929272 Chr.10: 86658182-86658182 on Build GRCh38
rs182222074 Chr.10: 86658004-86658004 on Build GRCh38
rs1003942522 Chr.10: 86658114-86658114 on Build GRCh38
rs759848344 Chr.10: 86658164-86658164 on Build GRCh38
rs991000251 Chr.10: 86658149-86658149 on Build GRCh38
rs752286086 Chr.10: 86658178-86658178 on Build GRCh38
rs977850019 Chr.10: 86658184-86658184 on Build GRCh38
rs543587644 Chr.10: 86658045-86658045 on Build GRCh38
rs746925129 Chr.10: 86658044-86658050 on Build GRCh38
rs1022425231 Chr.10: 86658179-86658179 on Build GRCh38
rs765874223 Chr.10: 86658014-86658014 on Build GRCh38
rs775836075 Chr.10: 86657997-86657997 on Build GRCh38
rs755583201 Chr.10: 86658053-86658053 on Build GRCh38
rs887639144 Chr.10: 86658106-86658106 on Build GRCh38
rs201523315 Chr.10: 86658117-86658117 on Build GRCh38
rs763365909 Chr.10: 86658116-86658116 on Build GRCh38

View Full Product Details