Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 65729513-65729752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSN
Entrez Gene ID
4478
Gene Name
moesin
Gene Aliases
HEL70
UniGene
Hs.713679
Gene Chromosome Location
Chr.X: 65588382-65741931 on Build GRCh38
Transcripts
NM_002444.2 XM_005262269.2 XM_011530959.1 XM_017029545.1 XM_017029546.1

SNP Detail

SNP ID Chromosome Location
rs759053380 Chr.X: 65729662-65729662 on Build GRCh38
rs753903006 Chr.X: 65729698-65729698 on Build GRCh38
rs762194075 Chr.X: 65729647-65729647 on Build GRCh38
rs188604598 Chr.X: 65729608-65729608 on Build GRCh38
rs982521952 Chr.X: 65729598-65729598 on Build GRCh38
rs745873198 Chr.X: 65729707-65729707 on Build GRCh38
rs770945316 Chr.X: 65729544-65729544 on Build GRCh38
rs746950857 Chr.X: 65729543-65729543 on Build GRCh38
rs753331094 Chr.X: 65729658-65729658 on Build GRCh38
rs140037207 Chr.X: 65729688-65729688 on Build GRCh38
rs768010742 Chr.X: 65729715-65729715 on Build GRCh38
rs746005784 Chr.X: 65729599-65729599 on Build GRCh38
rs757131925 Chr.X: 65729690-65729690 on Build GRCh38
rs146058241 Chr.X: 65729674-65729674 on Build GRCh38
rs199526068 Chr.X: 65729675-65729675 on Build GRCh38
rs886151880 Chr.X: 65729602-65729602 on Build GRCh38
rs756272887 Chr.X: 65729722-65729722 on Build GRCh38
rs774881056 Chr.X: 65729644-65729644 on Build GRCh38
rs368950777 Chr.X: 65729689-65729689 on Build GRCh38
rs763348254 Chr.X: 65729617-65729617 on Build GRCh38
rs775857259 Chr.X: 65729609-65729609 on Build GRCh38
rs781113630 Chr.X: 65729563-65729563 on Build GRCh38
rs760751746 Chr.X: 65729626-65729626 on Build GRCh38
rs758172397 Chr.X: 65729676-65729676 on Build GRCh38
rs768921044 Chr.X: 65729618-65729618 on Build GRCh38
rs768117369 Chr.X: 65729651-65729651 on Build GRCh38

View Full Product Details