Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41476987-41477256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT35
Entrez Gene ID
3886
Gene Name
keratin 35
Gene Aliases
HA5, Ha-5, KRTHA5, hHa5
UniGene
Hs.73082
Gene Chromosome Location
Chr.17: 41476689-41482743 on Build GRCh38
Transcripts
NM_002280.4 XM_011524794.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
124027 c.1310C>T p.A437V Chr.17: 41477114-41477114 on Build GRCh38
3932593 c.1359T>C p.G453G Chr.17: 41477065-41477065 on Build GRCh38
6698008 c.1221-5_1221-4insT p.? Chr.17: 41477207-41477208 on Build GRCh38
3770487 c.1307C>A p.A436E Chr.17: 41477117-41477117 on Build GRCh38
108487 c.1225C>T p.P409S Chr.17: 41477199-41477199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369127524 Chr.17: 41477138-41477138 on Build GRCh38
rs760987949 Chr.17: 41477101-41477101 on Build GRCh38
rs779922267 Chr.17: 41477131-41477131 on Build GRCh38
rs777055054 Chr.17: 41477159-41477159 on Build GRCh38
rs377487322 Chr.17: 41477067-41477067 on Build GRCh38
rs577006980 Chr.17: 41477044-41477044 on Build GRCh38
rs12451652 Chr.17: 41477102-41477102 on Build GRCh38
rs769662906 Chr.17: 41477093-41477093 on Build GRCh38
rs371696778 Chr.17: 41477213-41477213 on Build GRCh38
rs201141610 Chr.17: 41477051-41477051 on Build GRCh38
rs773598088 Chr.17: 41477030-41477030 on Build GRCh38
rs759031922 Chr.17: 41477064-41477064 on Build GRCh38
rs777526892 Chr.17: 41477069-41477069 on Build GRCh38
rs762039687 Chr.17: 41477016-41477016 on Build GRCh38
rs763886413 Chr.17: 41477124-41477124 on Build GRCh38
rs200459327 Chr.17: 41477122-41477122 on Build GRCh38
rs368599803 Chr.17: 41477098-41477098 on Build GRCh38
rs754337552 Chr.17: 41477059-41477059 on Build GRCh38
rs761850237 Chr.17: 41477215-41477215 on Build GRCh38
rs777163609 Chr.17: 41477207-41477208 on Build GRCh38
rs868681711 Chr.17: 41477079-41477079 on Build GRCh38
rs769323096 Chr.17: 41477029-41477029 on Build GRCh38
rs778969320 Chr.17: 41477094-41477094 on Build GRCh38
rs751191773 Chr.17: 41477126-41477126 on Build GRCh38
rs201907377 Chr.17: 41477127-41477127 on Build GRCh38
rs938522136 Chr.17: 41477181-41477181 on Build GRCh38
rs773636138 Chr.17: 41477093-41477093 on Build GRCh38
rs761533012 Chr.17: 41477209-41477209 on Build GRCh38
rs770997785 Chr.17: 41477150-41477150 on Build GRCh38
rs776117115 Chr.17: 41477175-41477175 on Build GRCh38
rs750259022 Chr.17: 41477226-41477226 on Build GRCh38
rs892754570 Chr.17: 41477074-41477074 on Build GRCh38
rs764539162 Chr.17: 41477189-41477189 on Build GRCh38
rs2239711 Chr.17: 41477065-41477065 on Build GRCh38
rs752286561 Chr.17: 41477119-41477119 on Build GRCh38
rs748278996 Chr.17: 41477132-41477132 on Build GRCh38
rs370694468 Chr.17: 41477089-41477089 on Build GRCh38
rs774779421 Chr.17: 41477112-41477112 on Build GRCh38
rs370552121 Chr.17: 41477052-41477052 on Build GRCh38
rs776010559 Chr.17: 41477154-41477154 on Build GRCh38
rs1001062970 Chr.17: 41477063-41477063 on Build GRCh38
rs2071601 Chr.17: 41477097-41477097 on Build GRCh38
rs746178649 Chr.17: 41477161-41477161 on Build GRCh38
rs770637190 Chr.17: 41477144-41477145 on Build GRCh38
rs757052525 Chr.17: 41477087-41477087 on Build GRCh38
rs751331700 Chr.17: 41477084-41477084 on Build GRCh38
rs758128272 Chr.17: 41477068-41477068 on Build GRCh38
rs762590390 Chr.17: 41477198-41477198 on Build GRCh38
rs777060154 Chr.17: 41477111-41477111 on Build GRCh38
rs773678576 Chr.17: 41477203-41477203 on Build GRCh38
rs772215595 Chr.17: 41477017-41477017 on Build GRCh38
rs150062210 Chr.17: 41477199-41477199 on Build GRCh38
rs532340423 Chr.17: 41477222-41477222 on Build GRCh38
rs747238448 Chr.17: 41477144-41477144 on Build GRCh38
rs765925126 Chr.17: 41477114-41477114 on Build GRCh38
rs367703815 Chr.17: 41477128-41477128 on Build GRCh38
rs374763543 Chr.17: 41477137-41477137 on Build GRCh38
rs772795562 Chr.17: 41477043-41477043 on Build GRCh38
rs571605941 Chr.17: 41477162-41477162 on Build GRCh38
rs761072193 Chr.17: 41477159-41477160 on Build GRCh38
rs759219201 Chr.17: 41477180-41477180 on Build GRCh38

View Full Product Details