Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122621257-122621530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARP15
Entrez Gene ID
165631
Gene Name
poly(ADP-ribose) polymerase family member 15
Gene Aliases
ARTD7, BAL3, pART7
UniGene
Hs.120250
Gene Chromosome Location
Chr.3: 122575914-122639047 on Build GRCh38
Transcripts
NM_001113523.2 NM_001308320.1 NM_001308321.1 NM_152615.2 XM_005247159.3 XM_005247160.3 XM_011512475.2 XM_011512476.2 XM_011512477.2 XM_011512478.2 XM_011512479.2 XM_017005791.1 XM_017005792.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1683952 c.1146delA p.D383fs*10 Chr.3: 122621526-122621526 on Build GRCh38
1693326 c.1141G>A p.G381R Chr.3: 122621521-122621521 on Build GRCh38
5355022 c.362-10delT p.? Chr.3: 122621434-122621434 on Build GRCh38
1037189 c.386T>C p.I129T Chr.3: 122621468-122621468 on Build GRCh38
6737880 c.1113G>T p.K371N Chr.3: 122621493-122621493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760995919 Chr.3: 122621500-122621500 on Build GRCh38
rs748166367 Chr.3: 122621489-122621489 on Build GRCh38
rs142282414 Chr.3: 122621503-122621503 on Build GRCh38
rs776496069 Chr.3: 122621506-122621506 on Build GRCh38
rs758284154 Chr.3: 122621446-122621446 on Build GRCh38
rs960285316 Chr.3: 122621377-122621377 on Build GRCh38
rs773728217 Chr.3: 122621493-122621493 on Build GRCh38
rs752190639 Chr.3: 122621438-122621438 on Build GRCh38
rs113970648 Chr.3: 122621475-122621475 on Build GRCh38
rs754596056 Chr.3: 122621395-122621395 on Build GRCh38
rs750452621 Chr.3: 122621417-122621417 on Build GRCh38
rs749495736 Chr.3: 122621476-122621476 on Build GRCh38
rs147746757 Chr.3: 122621463-122621463 on Build GRCh38
rs112470875 Chr.3: 122621326-122621326 on Build GRCh38
rs569515589 Chr.3: 122621325-122621325 on Build GRCh38
rs959004387 Chr.3: 122621347-122621347 on Build GRCh38
rs764836761 Chr.3: 122621405-122621405 on Build GRCh38
rs768787452 Chr.3: 122621478-122621478 on Build GRCh38
rs774479852 Chr.3: 122621443-122621445 on Build GRCh38
rs555860752 Chr.3: 122621287-122621287 on Build GRCh38
rs772478731 Chr.3: 122621492-122621492 on Build GRCh38
rs148790215 Chr.3: 122621474-122621474 on Build GRCh38
rs917806006 Chr.3: 122621358-122621358 on Build GRCh38
rs774417171 Chr.3: 122621481-122621481 on Build GRCh38
rs973100202 Chr.3: 122621425-122621425 on Build GRCh38
rs878931909 Chr.3: 122621385-122621386 on Build GRCh38
rs746815603 Chr.3: 122621458-122621458 on Build GRCh38
rs555094510 Chr.3: 122621354-122621354 on Build GRCh38
rs1032119607 Chr.3: 122621336-122621336 on Build GRCh38

View Full Product Details