Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 106363809-106364064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR3B
Entrez Gene ID
55703
Gene Name
RNA polymerase III subunit B
Gene Aliases
C128, HLD8, INMAP, RPC2
UniGene
Hs.610795
Gene Chromosome Location
Chr.12: 106357658-106510198 on Build GRCh38
Transcripts
NM_001160708.1 NM_018082.5 XM_017019621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6425137 c.84G>T p.R28S Chr.12: 106363881-106363881 on Build GRCh38
6849140 c.81G>T p.W27C Chr.12: 106363878-106363878 on Build GRCh38
81603 c.87G>T p.L29L Chr.12: 106363884-106363884 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs377362682 Chr.12: 106363931-106363931 on Build GRCh38
rs894802718 Chr.12: 106363929-106363929 on Build GRCh38
rs778246427 Chr.12: 106363921-106363921 on Build GRCh38
rs1011986009 Chr.12: 106363953-106363953 on Build GRCh38
rs991414901 Chr.12: 106363997-106363997 on Build GRCh38
rs749701042 Chr.12: 106363907-106363907 on Build GRCh38
rs865872026 Chr.12: 106363912-106363912 on Build GRCh38
rs140678013 Chr.12: 106363848-106363850 on Build GRCh38
rs898209969 Chr.12: 106364016-106364016 on Build GRCh38
rs775469366 Chr.12: 106363939-106363939 on Build GRCh38
rs781559477 Chr.12: 106363846-106363846 on Build GRCh38
rs958574829 Chr.12: 106363996-106363996 on Build GRCh38
rs78667485 Chr.12: 106363915-106363915 on Build GRCh38
rs779537305 Chr.12: 106363882-106363882 on Build GRCh38
rs761129848 Chr.12: 106364032-106364032 on Build GRCh38
rs879014204 Chr.12: 106363918-106363918 on Build GRCh38
rs773294868 Chr.12: 106364018-106364018 on Build GRCh38
rs778337028 Chr.12: 106363885-106363885 on Build GRCh38
rs914510226 Chr.12: 106364004-106364004 on Build GRCh38
rs755150098 Chr.12: 106363838-106363838 on Build GRCh38
rs567745261 Chr.12: 106363977-106363977 on Build GRCh38
rs753038075 Chr.12: 106363850-106363850 on Build GRCh38
rs770481246 Chr.12: 106363916-106363916 on Build GRCh38
rs772074660 Chr.12: 106363867-106363867 on Build GRCh38

View Full Product Details