Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 150582810-150583081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRBA
Entrez Gene ID
987
Gene Name
LPS responsive beige-like anchor protein
Gene Aliases
BGL, CDC4L, CVID8, LAB300, LBA
UniGene
Hs.480938
Gene Chromosome Location
Chr.4: 150264515-151015725 on Build GRCh38
Transcripts
NM_001199282.2 NM_006726.4 XM_005263372.3 XM_005263373.2 XM_005263374.3 XM_005263375.3 XM_011532434.2 XM_017008872.1 NM_006439.4

Gene Symbol
MAB21L2
Entrez Gene ID
10586
Gene Name
mab-21 like 2
Gene Aliases
MCOPS14
UniGene
Hs.584852
Gene Chromosome Location
Chr.4: 150581925-150584693 on Build GRCh38
Transcripts
NM_001199282.2 NM_006726.4 XM_005263372.3 XM_005263373.2 XM_005263374.3 XM_005263375.3 XM_011532434.2 XM_017008872.1 NM_006439.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5502908 c.22C>A p.L8M Chr.4: 150583051-150583051 on Build GRCh38
7277737 c.10G>A p.A4T Chr.4: 150583039-150583039 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371518534 Chr.4: 150582994-150582994 on Build GRCh38
rs560611550 Chr.4: 150582867-150582867 on Build GRCh38
rs1034382910 Chr.4: 150582940-150582940 on Build GRCh38
rs1011556096 Chr.4: 150582863-150582863 on Build GRCh38
rs1019547955 Chr.4: 150582901-150582901 on Build GRCh38
rs895369184 Chr.4: 150582990-150582990 on Build GRCh38
rs762352154 Chr.4: 150583007-150583007 on Build GRCh38
rs965156364 Chr.4: 150582962-150582962 on Build GRCh38
rs748833323 Chr.4: 150582837-150582838 on Build GRCh38
rs940078616 Chr.4: 150582891-150582891 on Build GRCh38
rs751766566 Chr.4: 150583050-150583050 on Build GRCh38
rs762457430 Chr.4: 150582919-150582919 on Build GRCh38
rs115316565 Chr.4: 150582860-150582860 on Build GRCh38
rs878922026 Chr.4: 150582932-150582932 on Build GRCh38
rs995515298 Chr.4: 150582924-150582924 on Build GRCh38
rs933156506 Chr.4: 150582847-150582847 on Build GRCh38
rs889700621 Chr.4: 150582896-150582896 on Build GRCh38
rs1043487616 Chr.4: 150582870-150582870 on Build GRCh38
rs750545900 Chr.4: 150583014-150583014 on Build GRCh38
rs200992768 Chr.4: 150583039-150583039 on Build GRCh38
rs549286219 Chr.4: 150582897-150582897 on Build GRCh38
rs564828987 Chr.4: 150582842-150582842 on Build GRCh38
rs993992427 Chr.4: 150582983-150582983 on Build GRCh38
rs569029146 Chr.4: 150582921-150582921 on Build GRCh38
rs761139412 Chr.4: 150582979-150582979 on Build GRCh38
rs529523735 Chr.4: 150582877-150582877 on Build GRCh38
rs547315945 Chr.4: 150582864-150582864 on Build GRCh38
rs770495370 Chr.4: 150582996-150582996 on Build GRCh38
rs375783917 Chr.4: 150583013-150583013 on Build GRCh38

View Full Product Details