Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 98041764-98042019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL30
Entrez Gene ID
6156
Gene Name
ribosomal protein L30
Gene Aliases
L30
UniGene
Hs.400295
Gene Chromosome Location
Chr.8: 98041710-98045590 on Build GRCh38
Transcripts
NM_000989.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5830926 c.299-3delT p.? Chr.8: 98041853-98041853 on Build GRCh38
3395464 c.336T>C p.T112T Chr.8: 98041813-98041813 on Build GRCh38
5748374 c.301G>T p.D101Y Chr.8: 98041848-98041848 on Build GRCh38
6769221 c.299-3_299-2insT p.? Chr.8: 98041852-98041853 on Build GRCh38
7291883 c.299-10_299-9insT p.? Chr.8: 98041859-98041860 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776381480 Chr.8: 98041898-98041899 on Build GRCh38
rs909721328 Chr.8: 98041962-98041962 on Build GRCh38
rs745534956 Chr.8: 98041899-98041900 on Build GRCh38
rs780519383 Chr.8: 98041882-98041882 on Build GRCh38
rs768960745 Chr.8: 98041899-98041899 on Build GRCh38
rs879141554 Chr.8: 98041856-98041856 on Build GRCh38
rs1804308 Chr.8: 98041826-98041826 on Build GRCh38
rs556317748 Chr.8: 98041987-98041987 on Build GRCh38
rs374951962 Chr.8: 98041861-98041861 on Build GRCh38
rs536581333 Chr.8: 98041942-98041942 on Build GRCh38
rs1037764177 Chr.8: 98041979-98041980 on Build GRCh38
rs770464342 Chr.8: 98041866-98041869 on Build GRCh38
rs879210878 Chr.8: 98041855-98041855 on Build GRCh38
rs878894467 Chr.8: 98041854-98041854 on Build GRCh38
rs367653698 Chr.8: 98041873-98041873 on Build GRCh38
rs879097318 Chr.8: 98041852-98041853 on Build GRCh38
rs746592489 Chr.8: 98041866-98041871 on Build GRCh38
rs372903661 Chr.8: 98041878-98041878 on Build GRCh38
rs879191763 Chr.8: 98041857-98041857 on Build GRCh38
rs925258860 Chr.8: 98041945-98041945 on Build GRCh38
rs958046341 Chr.8: 98041983-98041983 on Build GRCh38
rs752161188 Chr.8: 98041822-98041822 on Build GRCh38
rs878968544 Chr.8: 98041853-98041853 on Build GRCh38
rs762871107 Chr.8: 98041940-98041940 on Build GRCh38
rs141159921 Chr.8: 98041911-98041911 on Build GRCh38
rs772584326 Chr.8: 98041865-98041866 on Build GRCh38
rs139559856 Chr.8: 98041814-98041814 on Build GRCh38
rs769608930 Chr.8: 98041903-98041903 on Build GRCh38
rs941159764 Chr.8: 98041974-98041974 on Build GRCh38
rs201910608 Chr.8: 98041816-98041816 on Build GRCh38
rs747171428 Chr.8: 98041897-98041897 on Build GRCh38
rs780648509 Chr.8: 98041792-98041792 on Build GRCh38
rs966594503 Chr.8: 98041895-98041895 on Build GRCh38
rs779329839 Chr.8: 98041862-98041862 on Build GRCh38

View Full Product Details