Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46692343-46692594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ORC6
Entrez Gene ID
23594
Gene Name
origin recognition complex subunit 6
Gene Aliases
ORC6L
UniGene
Hs.49760
Gene Chromosome Location
Chr.16: 46689646-46698394 on Build GRCh38
Transcripts
NM_014321.3 NR_037620.1 XM_011522978.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6002205 c.196-2A>G p.? Chr.16: 46692380-46692380 on Build GRCh38
7114461 c.296G>C p.R99T Chr.16: 46692482-46692482 on Build GRCh38
970768 c.196-1G>T p.? Chr.16: 46692381-46692381 on Build GRCh38
4542815 c.325G>A p.E109K Chr.16: 46692511-46692511 on Build GRCh38
3888532 c.359G>A p.S120N Chr.16: 46692545-46692545 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765526371 Chr.16: 46692435-46692435 on Build GRCh38
rs200089121 Chr.16: 46692421-46692421 on Build GRCh38
rs138692254 Chr.16: 46692508-46692508 on Build GRCh38
rs752050185 Chr.16: 46692486-46692486 on Build GRCh38
rs746948898 Chr.16: 46692517-46692517 on Build GRCh38
rs142697731 Chr.16: 46692509-46692509 on Build GRCh38
rs143913118 Chr.16: 46692526-46692526 on Build GRCh38
rs138076975 Chr.16: 46692393-46692393 on Build GRCh38
rs786205258 Chr.16: 46692443-46692444 on Build GRCh38
rs755612392 Chr.16: 46692487-46692487 on Build GRCh38
rs779455633 Chr.16: 46692495-46692495 on Build GRCh38
rs775741529 Chr.16: 46692553-46692553 on Build GRCh38
rs770990429 Chr.16: 46692525-46692525 on Build GRCh38
rs748032792 Chr.16: 46692423-46692423 on Build GRCh38
rs759732425 Chr.16: 46692547-46692547 on Build GRCh38
rs937842346 Chr.16: 46692472-46692472 on Build GRCh38
rs936646606 Chr.16: 46692499-46692508 on Build GRCh38
rs762477641 Chr.16: 46692451-46692451 on Build GRCh38
rs372381137 Chr.16: 46692569-46692569 on Build GRCh38
rs768450500 Chr.16: 46692382-46692382 on Build GRCh38
rs758826283 Chr.16: 46692467-46692467 on Build GRCh38
rs776871410 Chr.16: 46692424-46692424 on Build GRCh38
rs138295997 Chr.16: 46692478-46692478 on Build GRCh38
rs143750053 Chr.16: 46692429-46692429 on Build GRCh38
rs773340569 Chr.16: 46692413-46692413 on Build GRCh38
rs763277494 Chr.16: 46692559-46692559 on Build GRCh38
rs778774952 Chr.16: 46692505-46692505 on Build GRCh38
rs748927298 Chr.16: 46692381-46692381 on Build GRCh38
rs765437962 Chr.16: 46692552-46692552 on Build GRCh38
rs375110765 Chr.16: 46692570-46692570 on Build GRCh38
rs766253178 Chr.16: 46692499-46692499 on Build GRCh38
rs747944499 Chr.16: 46692506-46692506 on Build GRCh38
rs748032369 Chr.16: 46692384-46692384 on Build GRCh38
rs147215492 Chr.16: 46692462-46692462 on Build GRCh38
rs754684270 Chr.16: 46692501-46692501 on Build GRCh38
rs1046633319 Chr.16: 46692540-46692540 on Build GRCh38
rs751458368 Chr.16: 46692551-46692551 on Build GRCh38

View Full Product Details