Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 57052612-57052837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1586
Entrez Gene ID
57691
Gene Name
KIAA1586
Gene Aliases
-
UniGene
Hs.709521
Gene Chromosome Location
Chr.6: 57046532-57055739 on Build GRCh38
Transcripts
NM_001286274.1 NM_001286275.1 NM_001286276.1 NM_020931.3 XM_011514767.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3875006 c.212G>A p.R71Q Chr.6: 57052711-57052711 on Build GRCh38
5912446 c.197C>T p.P66L Chr.6: 57052696-57052696 on Build GRCh38
484267 c.226T>A p.L76M Chr.6: 57052725-57052725 on Build GRCh38
3762147 c.241G>A p.V81M Chr.6: 57052740-57052740 on Build GRCh38
3875008 c.325G>A p.E109K Chr.6: 57052824-57052824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761463864 Chr.6: 57052735-57052735 on Build GRCh38
rs764785600 Chr.6: 57052645-57052645 on Build GRCh38
rs754751249 Chr.6: 57052760-57052760 on Build GRCh38
rs61741654 Chr.6: 57052710-57052710 on Build GRCh38
rs777762311 Chr.6: 57052707-57052707 on Build GRCh38
rs780981237 Chr.6: 57052772-57052772 on Build GRCh38
rs942895242 Chr.6: 57052800-57052800 on Build GRCh38
rs748351972 Chr.6: 57052696-57052696 on Build GRCh38
rs199675919 Chr.6: 57052657-57052657 on Build GRCh38
rs373034964 Chr.6: 57052778-57052778 on Build GRCh38
rs762597758 Chr.6: 57052759-57052760 on Build GRCh38
rs779313975 Chr.6: 57052805-57052805 on Build GRCh38
rs750008823 Chr.6: 57052736-57052736 on Build GRCh38
rs371267376 Chr.6: 57052751-57052751 on Build GRCh38
rs766456500 Chr.6: 57052748-57052748 on Build GRCh38
rs749411448 Chr.6: 57052785-57052785 on Build GRCh38
rs534677385 Chr.6: 57052808-57052808 on Build GRCh38
rs780123680 Chr.6: 57052668-57052668 on Build GRCh38
rs36113897 Chr.6: 57052722-57052722 on Build GRCh38
rs752539903 Chr.6: 57052688-57052689 on Build GRCh38
rs774891326 Chr.6: 57052715-57052715 on Build GRCh38
rs757287899 Chr.6: 57052792-57052792 on Build GRCh38
rs570836745 Chr.6: 57052755-57052755 on Build GRCh38
rs754766714 Chr.6: 57052684-57052684 on Build GRCh38
rs751356155 Chr.6: 57052677-57052677 on Build GRCh38
rs781300637 Chr.6: 57052692-57052692 on Build GRCh38
rs750393262 Chr.6: 57052651-57052651 on Build GRCh38
rs776165556 Chr.6: 57052727-57052727 on Build GRCh38
rs577941240 Chr.6: 57052715-57052715 on Build GRCh38
rs147392367 Chr.6: 57052711-57052711 on Build GRCh38
rs765061638 Chr.6: 57052668-57052669 on Build GRCh38
rs769875082 Chr.6: 57052700-57052700 on Build GRCh38
rs991372120 Chr.6: 57052656-57052656 on Build GRCh38
rs912687483 Chr.6: 57052799-57052799 on Build GRCh38
rs6926980 Chr.6: 57052740-57052740 on Build GRCh38
rs777951207 Chr.6: 57052780-57052780 on Build GRCh38
rs267601092 Chr.6: 57052695-57052695 on Build GRCh38
rs759849940 Chr.6: 57052716-57052716 on Build GRCh38

View Full Product Details