Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 39461987-39462240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIAS
Entrez Gene ID
11019
Gene Name
lipoic acid synthetase
Gene Aliases
HGCLAS, HUSSY-01, LAS, LIP1, LS, PDHLD
UniGene
Hs.550502
Gene Chromosome Location
Chr.4: 39459022-39477653 on Build GRCh38
Transcripts
NM_001278590.1 NM_001278591.1 NM_001278592.1 NM_006859.3 NM_194451.2 XM_006713990.3 XM_017007665.1 XM_017007666.1 XM_017007667.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4611139 c.262delA p.N89fs*5 Chr.4: 39462239-39462239 on Build GRCh38
3732438 c.219-2A>G p.? Chr.4: 39462194-39462194 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749231188 Chr.4: 39462155-39462155 on Build GRCh38
rs760630502 Chr.4: 39462081-39462081 on Build GRCh38
rs1028853082 Chr.4: 39462121-39462121 on Build GRCh38
rs968048398 Chr.4: 39462096-39462096 on Build GRCh38
rs761462392 Chr.4: 39462169-39462169 on Build GRCh38
rs771651616 Chr.4: 39462180-39462180 on Build GRCh38
rs773366111 Chr.4: 39462070-39462070 on Build GRCh38
rs979822762 Chr.4: 39462099-39462099 on Build GRCh38
rs375322646 Chr.4: 39462159-39462159 on Build GRCh38
rs760903814 Chr.4: 39462196-39462196 on Build GRCh38
rs373474311 Chr.4: 39462144-39462144 on Build GRCh38
rs766730917 Chr.4: 39462211-39462211 on Build GRCh38
rs1021382881 Chr.4: 39462039-39462039 on Build GRCh38
rs569057005 Chr.4: 39462161-39462161 on Build GRCh38
rs991992347 Chr.4: 39462154-39462154 on Build GRCh38
rs908541464 Chr.4: 39462171-39462171 on Build GRCh38
rs536239528 Chr.4: 39462097-39462097 on Build GRCh38
rs183098295 Chr.4: 39462181-39462181 on Build GRCh38
rs368755029 Chr.4: 39462116-39462116 on Build GRCh38
rs549290648 Chr.4: 39462142-39462142 on Build GRCh38

View Full Product Details