Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100017681-100017908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35A3
Entrez Gene ID
23443
Gene Name
solute carrier family 35 member A3
Gene Aliases
AMRS
UniGene
Hs.448979
Gene Chromosome Location
Chr.1: 99969367-100026979 on Build GRCh38
Transcripts
NM_001271684.1 NM_001271685.1 NM_012243.2 XM_005270691.4 XM_011541135.2 XM_011541136.2 XM_011541137.2 XM_011541138.2 XM_017000869.1 XM_017000870.1 XM_017000871.1 XM_017000872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7282910 c.822C>A p.T274T Chr.1: 100017750-100017750 on Build GRCh38
1688113 c.763_764GG>AA p.G255N Chr.1: 100017691-100017692 on Build GRCh38
308569 c.842C>T p.S281L Chr.1: 100017770-100017770 on Build GRCh38
4759366 c.857A>G p.Y286C Chr.1: 100017785-100017785 on Build GRCh38
4656086 c.880G>A p.A294T Chr.1: 100017682-100017682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750680399 Chr.1: 100017726-100017726 on Build GRCh38
rs370295328 Chr.1: 100017838-100017838 on Build GRCh38
rs748356892 Chr.1: 100017735-100017735 on Build GRCh38
rs778337201 Chr.1: 100017833-100017834 on Build GRCh38
rs749734512 Chr.1: 100017759-100017759 on Build GRCh38
rs755533972 Chr.1: 100017766-100017766 on Build GRCh38
rs756106534 Chr.1: 100017735-100017735 on Build GRCh38
rs775331346 Chr.1: 100017858-100017858 on Build GRCh38
rs371935609 Chr.1: 100017778-100017778 on Build GRCh38
rs747536139 Chr.1: 100017753-100017754 on Build GRCh38
rs141952252 Chr.1: 100017814-100017814 on Build GRCh38
rs552844460 Chr.1: 100017824-100017828 on Build GRCh38
rs1044884618 Chr.1: 100017871-100017871 on Build GRCh38
rs1023907141 Chr.1: 100017868-100017868 on Build GRCh38
rs572503397 Chr.1: 100017849-100017849 on Build GRCh38
rs772703781 Chr.1: 100017823-100017823 on Build GRCh38
rs767680002 Chr.1: 100017719-100017719 on Build GRCh38
rs776340329 Chr.1: 100017839-100017839 on Build GRCh38
rs12123408 Chr.1: 100017836-100017836 on Build GRCh38
rs779241992 Chr.1: 100017775-100017775 on Build GRCh38
rs780108041 Chr.1: 100017758-100017758 on Build GRCh38
rs541734968 Chr.1: 100017874-100017874 on Build GRCh38
rs759544206 Chr.1: 100017845-100017845 on Build GRCh38
rs907333653 Chr.1: 100017878-100017878 on Build GRCh38
rs960736132 Chr.1: 100017773-100017773 on Build GRCh38
rs757468494 Chr.1: 100017716-100017716 on Build GRCh38

View Full Product Details