Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 79520817-79521060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDFIP2
Entrez Gene ID
54602
Gene Name
Nedd4 family interacting protein 2
Gene Aliases
N4WBP5A
UniGene
Hs.525093
Gene Chromosome Location
Chr.13: 79480722-79556077 on Build GRCh38
Transcripts
NM_001161407.1 NM_019080.2 XM_006719840.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
71800 c.459T>A p.S153R Chr.13: 79520947-79520947 on Build GRCh38
1367949 c.390G>A p.P130P Chr.13: 79520878-79520878 on Build GRCh38
7230543 c.358A>G p.I120V Chr.13: 79520846-79520846 on Build GRCh38
4147777 c.407C>T p.A136V Chr.13: 79520895-79520895 on Build GRCh38
227859 c.389C>T p.P130L Chr.13: 79520877-79520877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548758177 Chr.13: 79520882-79520882 on Build GRCh38
rs762057502 Chr.13: 79521005-79521006 on Build GRCh38
rs751254644 Chr.13: 79520898-79520898 on Build GRCh38
rs757951274 Chr.13: 79520890-79520890 on Build GRCh38
rs756884977 Chr.13: 79520915-79520915 on Build GRCh38
rs775531268 Chr.13: 79521014-79521014 on Build GRCh38
rs780458623 Chr.13: 79520921-79520921 on Build GRCh38
rs755234462 Chr.13: 79520938-79520938 on Build GRCh38
rs58406632 Chr.13: 79521022-79521022 on Build GRCh38
rs745887981 Chr.13: 79521004-79521004 on Build GRCh38
rs776355050 Chr.13: 79520994-79520994 on Build GRCh38
rs61735407 Chr.13: 79520943-79520943 on Build GRCh38
rs59405124 Chr.13: 79521020-79521021 on Build GRCh38
rs764892636 Chr.13: 79521008-79521008 on Build GRCh38
rs996269160 Chr.13: 79520976-79520976 on Build GRCh38
rs747282493 Chr.13: 79520966-79520966 on Build GRCh38
rs774365204 Chr.13: 79520855-79520855 on Build GRCh38
rs755665561 Chr.13: 79521009-79521010 on Build GRCh38
rs759379013 Chr.13: 79521001-79521001 on Build GRCh38
rs368345034 Chr.13: 79520878-79520878 on Build GRCh38
rs371388745 Chr.13: 79520896-79520896 on Build GRCh38
rs1043805159 Chr.13: 79520986-79520986 on Build GRCh38
rs529804990 Chr.13: 79520924-79520924 on Build GRCh38
rs371917287 Chr.13: 79520948-79520948 on Build GRCh38
rs748552976 Chr.13: 79520951-79520951 on Build GRCh38
rs748973342 Chr.13: 79520885-79520885 on Build GRCh38
rs772217331 Chr.13: 79520859-79520859 on Build GRCh38
rs760936702 Chr.13: 79520864-79520864 on Build GRCh38
rs1047152716 Chr.13: 79520909-79520909 on Build GRCh38
rs773586282 Chr.13: 79520861-79520861 on Build GRCh38
rs373903171 Chr.13: 79520876-79520876 on Build GRCh38
rs749662530 Chr.13: 79520922-79520922 on Build GRCh38
rs766305137 Chr.13: 79521006-79521008 on Build GRCh38
rs532345659 Chr.13: 79520843-79520843 on Build GRCh38
rs546106538 Chr.13: 79520844-79520844 on Build GRCh38
rs763644072 Chr.13: 79521024-79521024 on Build GRCh38
rs371287967 Chr.13: 79520969-79520969 on Build GRCh38
rs55887763 Chr.13: 79520858-79520858 on Build GRCh38
rs146047464 Chr.13: 79520877-79520877 on Build GRCh38
rs773391730 Chr.13: 79520965-79520965 on Build GRCh38
rs908146057 Chr.13: 79520913-79520913 on Build GRCh38
rs144950614 Chr.13: 79520850-79520850 on Build GRCh38
rs769196655 Chr.13: 79520853-79520853 on Build GRCh38
rs573097381 Chr.13: 79521008-79521009 on Build GRCh38
rs759378523 Chr.13: 79521009-79521009 on Build GRCh38
rs772305104 Chr.13: 79520963-79520963 on Build GRCh38
rs752520267 Chr.13: 79520883-79520883 on Build GRCh38
rs11549502 Chr.13: 79520895-79520895 on Build GRCh38

View Full Product Details