Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49596393-49596628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR12
Entrez Gene ID
57479
Gene Name
proline rich 12
Gene Aliases
KIAA1205
UniGene
Hs.590971
Gene Chromosome Location
Chr.19: 49591177-49626439 on Build GRCh38
Transcripts
XM_017027031.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
713396 c.2216C>T p.P739L Chr.19: 49596551-49596551 on Build GRCh38
7251545 c.2131G>C p.D711H Chr.19: 49596466-49596466 on Build GRCh38
1395272 c.2179G>A p.E727K Chr.19: 49596514-49596514 on Build GRCh38
3536852 c.2127C>T p.S709S Chr.19: 49596462-49596462 on Build GRCh38
7239781 c.2116A>T p.T706S Chr.19: 49596451-49596451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372723162 Chr.19: 49596489-49596489 on Build GRCh38
rs199785002 Chr.19: 49596482-49596482 on Build GRCh38
rs748231541 Chr.19: 49596567-49596567 on Build GRCh38
rs557659919 Chr.19: 49596543-49596543 on Build GRCh38
rs777023768 Chr.19: 49596417-49596417 on Build GRCh38
rs774901947 Chr.19: 49596592-49596592 on Build GRCh38
rs774024771 Chr.19: 49596517-49596517 on Build GRCh38
rs373893428 Chr.19: 49596414-49596414 on Build GRCh38
rs766948254 Chr.19: 49596532-49596532 on Build GRCh38
rs775050452 Chr.19: 49596535-49596535 on Build GRCh38
rs759069592 Chr.19: 49596448-49596448 on Build GRCh38
rs938378766 Chr.19: 49596526-49596526 on Build GRCh38
rs759122948 Chr.19: 49596519-49596519 on Build GRCh38
rs752583300 Chr.19: 49596450-49596450 on Build GRCh38
rs756019654 Chr.19: 49596455-49596455 on Build GRCh38
rs980654778 Chr.19: 49596501-49596501 on Build GRCh38
rs773615736 Chr.19: 49596433-49596433 on Build GRCh38
rs549754880 Chr.19: 49596463-49596463 on Build GRCh38
rs771291544 Chr.19: 49596585-49596585 on Build GRCh38
rs543523020 Chr.19: 49596583-49596583 on Build GRCh38
rs756631811 Chr.19: 49596577-49596577 on Build GRCh38
rs755217328 Chr.19: 49596552-49596552 on Build GRCh38
rs745576901 Chr.19: 49596589-49596589 on Build GRCh38
rs375882447 Chr.19: 49596498-49596498 on Build GRCh38
rs375354908 Chr.19: 49596561-49596561 on Build GRCh38
rs762286566 Chr.19: 49596515-49596517 on Build GRCh38
rs963549226 Chr.19: 49596516-49596516 on Build GRCh38
rs372510933 Chr.19: 49596449-49596449 on Build GRCh38
rs372185474 Chr.19: 49596544-49596544 on Build GRCh38
rs368659022 Chr.19: 49596487-49596487 on Build GRCh38
rs753681228 Chr.19: 49596459-49596459 on Build GRCh38
rs765988941 Chr.19: 49596444-49596444 on Build GRCh38
rs554144415 Chr.19: 49596483-49596483 on Build GRCh38
rs762664639 Chr.19: 49596436-49596436 on Build GRCh38
rs770071734 Chr.19: 49596424-49596424 on Build GRCh38
rs771750435 Chr.19: 49596591-49596591 on Build GRCh38
rs568034897 Chr.19: 49596471-49596471 on Build GRCh38
rs780397535 Chr.19: 49596466-49596466 on Build GRCh38
rs369152968 Chr.19: 49596582-49596582 on Build GRCh38
rs774643093 Chr.19: 49596586-49596586 on Build GRCh38
rs762338141 Chr.19: 49596420-49596420 on Build GRCh38
rs753732058 Chr.19: 49596538-49596538 on Build GRCh38
rs770334014 Chr.19: 49596486-49596486 on Build GRCh38
rs927448221 Chr.19: 49596514-49596514 on Build GRCh38
rs747051347 Chr.19: 49596411-49596411 on Build GRCh38
rs531265357 Chr.19: 49596458-49596458 on Build GRCh38
rs888331179 Chr.19: 49596557-49596557 on Build GRCh38
rs189093151 Chr.19: 49596447-49596447 on Build GRCh38
rs1016076753 Chr.19: 49596504-49596504 on Build GRCh38
rs368869078 Chr.19: 49596536-49596536 on Build GRCh38

View Full Product Details