Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 84179753-84180006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAF1C
Entrez Gene ID
9013
Gene Name
TATA-box binding protein associated factor, RNA polymerase I subunit C
Gene Aliases
MGC:39976, SL1, TAFI110, TAFI95
UniGene
Hs.153022
Gene Chromosome Location
Chr.16: 84177847-84187070 on Build GRCh38
Transcripts
NM_001243156.1 NM_001243157.1 NM_001243158.1 NM_001243159.1 NM_001243160.1 NM_005679.3 NM_139353.2 XM_005256226.3 XM_005256227.3 XM_006721325.3 XM_006721326.3 XM_017023845.1 XM_017023846.1 XM_017023847.1 XM_017023848.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6839164 c.1745G>T p.G582V Chr.16: 84179806-84179806 on Build GRCh38
5559768 c.1725G>T p.L575F Chr.16: 84179826-84179826 on Build GRCh38
7227729 c.1748T>G p.L583R Chr.16: 84179803-84179803 on Build GRCh38
3755140 c.1723T>A p.L575M Chr.16: 84179828-84179828 on Build GRCh38
4828773 c.1766C>T p.S589L Chr.16: 84179785-84179785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753773560 Chr.16: 84179969-84179969 on Build GRCh38
rs775280278 Chr.16: 84179832-84179832 on Build GRCh38
rs770925328 Chr.16: 84179820-84179820 on Build GRCh38
rs751889922 Chr.16: 84179940-84179940 on Build GRCh38
rs778573041 Chr.16: 84179926-84179926 on Build GRCh38
rs779241189 Chr.16: 84179789-84179789 on Build GRCh38
rs374766366 Chr.16: 84179923-84179923 on Build GRCh38
rs781319630 Chr.16: 84179860-84179860 on Build GRCh38
rs766455723 Chr.16: 84179783-84179783 on Build GRCh38
rs530659290 Chr.16: 84179914-84179914 on Build GRCh38
rs754546154 Chr.16: 84179974-84179974 on Build GRCh38
rs570039965 Chr.16: 84179934-84179934 on Build GRCh38
rs377711443 Chr.16: 84179784-84179784 on Build GRCh38
rs200498274 Chr.16: 84179824-84179824 on Build GRCh38
rs746955225 Chr.16: 84179818-84179818 on Build GRCh38
rs548113450 Chr.16: 84179840-84179840 on Build GRCh38
rs527870933 Chr.16: 84179837-84179837 on Build GRCh38
rs764266270 Chr.16: 84179938-84179938 on Build GRCh38
rs748285352 Chr.16: 84179801-84179801 on Build GRCh38
rs766280271 Chr.16: 84179967-84179967 on Build GRCh38
rs760092034 Chr.16: 84179932-84179932 on Build GRCh38
rs769405301 Chr.16: 84179862-84179862 on Build GRCh38
rs776921037 Chr.16: 84179929-84179929 on Build GRCh38
rs186382482 Chr.16: 84179928-84179928 on Build GRCh38
rs757976739 Chr.16: 84179979-84179979 on Build GRCh38
rs745419883 Chr.16: 84179826-84179826 on Build GRCh38
rs768156867 Chr.16: 84179884-84179884 on Build GRCh38
rs201532742 Chr.16: 84179929-84179930 on Build GRCh38
rs777416461 Chr.16: 84179817-84179817 on Build GRCh38
rs368523970 Chr.16: 84179897-84179897 on Build GRCh38
rs748977427 Chr.16: 84179870-84179870 on Build GRCh38
rs4150172 Chr.16: 84179779-84179779 on Build GRCh38
rs577810185 Chr.16: 84179976-84179976 on Build GRCh38
rs767427337 Chr.16: 84179953-84179953 on Build GRCh38
rs539139538 Chr.16: 84179948-84179948 on Build GRCh38
rs371382879 Chr.16: 84179785-84179785 on Build GRCh38
rs562110474 Chr.16: 84179808-84179808 on Build GRCh38
rs1033727938 Chr.16: 84179949-84179949 on Build GRCh38
rs781123354 Chr.16: 84179911-84179911 on Build GRCh38
rs3217259 Chr.16: 84179935-84179936 on Build GRCh38
rs368174328 Chr.16: 84179886-84179886 on Build GRCh38
rs567843279 Chr.16: 84179900-84179900 on Build GRCh38
rs4150170 Chr.16: 84179833-84179833 on Build GRCh38
rs781642896 Chr.16: 84179986-84179986 on Build GRCh38
rs887331125 Chr.16: 84179919-84179919 on Build GRCh38
rs144584702 Chr.16: 84179966-84179966 on Build GRCh38
rs752651057 Chr.16: 84179892-84179892 on Build GRCh38
rs771250818 Chr.16: 84179888-84179888 on Build GRCh38
rs778087874 Chr.16: 84179781-84179781 on Build GRCh38
rs536685562 Chr.16: 84179915-84179915 on Build GRCh38
rs747810069 Chr.16: 84179978-84179978 on Build GRCh38
rs761072682 Chr.16: 84179887-84179887 on Build GRCh38
rs397840353 Chr.16: 84179938-84179939 on Build GRCh38
rs755123097 Chr.16: 84179841-84179841 on Build GRCh38
rs763748022 Chr.16: 84179936-84179937 on Build GRCh38
rs777714969 Chr.16: 84179849-84179849 on Build GRCh38
rs374279708 Chr.16: 84179782-84179782 on Build GRCh38
rs753019526 Chr.16: 84179843-84179843 on Build GRCh38
rs201138305 Chr.16: 84179778-84179778 on Build GRCh38
rs757096196 Chr.16: 84179859-84179859 on Build GRCh38
rs745797416 Chr.16: 84179861-84179861 on Build GRCh38
rs559053682 Chr.16: 84179954-84179954 on Build GRCh38
rs758489291 Chr.16: 84179845-84179845 on Build GRCh38
rs768186773 Chr.16: 84179924-84179924 on Build GRCh38
rs376946647 Chr.16: 84179835-84179835 on Build GRCh38
rs2230129 Chr.16: 84179828-84179828 on Build GRCh38
rs779377003 Chr.16: 84179863-84179863 on Build GRCh38
rs762983406 Chr.16: 84179935-84179935 on Build GRCh38
rs899997426 Chr.16: 84179909-84179909 on Build GRCh38
rs766819736 Chr.16: 84179838-84179838 on Build GRCh38
rs371858260 Chr.16: 84179889-84179889 on Build GRCh38
rs372288005 Chr.16: 84179910-84179910 on Build GRCh38

View Full Product Details