Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48806376-48806647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAT2
Entrez Gene ID
587
Gene Name
branched chain amino acid transaminase 2
Gene Aliases
BCAM, BCATM, BCT2, PP18
UniGene
Hs.512670
Gene Chromosome Location
Chr.19: 48795062-48811063 on Build GRCh38
Transcripts
NM_001164773.1 NM_001190.3 NM_001284325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3536666 c.294C>T p.S98S Chr.19: 48806523-48806523 on Build GRCh38
7076758 c.285C>G p.L95L Chr.19: 48806532-48806532 on Build GRCh38
7089279 c.172G>T p.G58W Chr.19: 48806645-48806645 on Build GRCh38
3422985 c.236G>A p.R79Q Chr.19: 48806581-48806581 on Build GRCh38
4924402 c.173G>C p.G58A Chr.19: 48806644-48806644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151164360 Chr.19: 48806616-48806616 on Build GRCh38
rs368226719 Chr.19: 48806469-48806469 on Build GRCh38
rs767818990 Chr.19: 48806511-48806511 on Build GRCh38
rs886710460 Chr.19: 48806581-48806581 on Build GRCh38
rs755321686 Chr.19: 48806506-48806506 on Build GRCh38
rs750358732 Chr.19: 48806601-48806601 on Build GRCh38
rs574280145 Chr.19: 48806550-48806550 on Build GRCh38
rs769538697 Chr.19: 48806474-48806474 on Build GRCh38
rs954008582 Chr.19: 48806407-48806407 on Build GRCh38
rs142099967 Chr.19: 48806597-48806597 on Build GRCh38
rs757196220 Chr.19: 48806547-48806547 on Build GRCh38
rs766500999 Chr.19: 48806566-48806566 on Build GRCh38
rs748821442 Chr.19: 48806540-48806540 on Build GRCh38
rs375689643 Chr.19: 48806507-48806507 on Build GRCh38
rs371062417 Chr.19: 48806503-48806503 on Build GRCh38
rs771881228 Chr.19: 48806555-48806555 on Build GRCh38
rs758302662 Chr.19: 48806609-48806609 on Build GRCh38
rs368301644 Chr.19: 48806552-48806552 on Build GRCh38
rs376551670 Chr.19: 48806589-48806589 on Build GRCh38
rs371450690 Chr.19: 48806588-48806588 on Build GRCh38
rs756816829 Chr.19: 48806590-48806590 on Build GRCh38
rs140816608 Chr.19: 48806557-48806557 on Build GRCh38
rs945249685 Chr.19: 48806500-48806500 on Build GRCh38
rs760073453 Chr.19: 48806568-48806568 on Build GRCh38
rs1021578203 Chr.19: 48806477-48806477 on Build GRCh38
rs117048185 Chr.19: 48806519-48806519 on Build GRCh38
rs968538707 Chr.19: 48806487-48806487 on Build GRCh38
rs767882479 Chr.19: 48806571-48806571 on Build GRCh38
rs766873285 Chr.19: 48806502-48806502 on Build GRCh38
rs779917358 Chr.19: 48806496-48806496 on Build GRCh38
rs770402005 Chr.19: 48806624-48806624 on Build GRCh38
rs868867197 Chr.19: 48806605-48806605 on Build GRCh38
rs201626538 Chr.19: 48806579-48806579 on Build GRCh38
rs775192346 Chr.19: 48806556-48806556 on Build GRCh38
rs368054122 Chr.19: 48806563-48806563 on Build GRCh38
rs376342351 Chr.19: 48806463-48806463 on Build GRCh38
rs753025021 Chr.19: 48806582-48806582 on Build GRCh38
rs752363280 Chr.19: 48806517-48806517 on Build GRCh38
rs768924723 Chr.19: 48806561-48806561 on Build GRCh38
rs1049112923 Chr.19: 48806553-48806553 on Build GRCh38
rs200449924 Chr.19: 48806499-48806499 on Build GRCh38
rs774816859 Chr.19: 48806567-48806567 on Build GRCh38
rs986610547 Chr.19: 48806424-48806424 on Build GRCh38

View Full Product Details