Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 69054956-69055221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPCN2
Entrez Gene ID
219931
Gene Name
two pore segment channel 2
Gene Aliases
SHEP10, TPC2
UniGene
Hs.131851
Gene Chromosome Location
Chr.11: 69048876-69090604 on Build GRCh38
Transcripts
NM_139075.3 XM_005273824.3 XM_005273826.3 XM_005273830.3 XM_005273831.3 XM_005273832.3 XM_011544802.2 XM_011544805.1 XM_011544806.1 XM_011544807.2 XM_011544808.2 XM_017017328.1 XM_017017329.1 XM_017017330.1 XM_017017331.1 XM_017017332.1 XM_017017333.1 XM_017017334.1 XM_017017335.1 XM_017017336.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5787097 c.258G>C p.L86F Chr.11: 69055181-69055181 on Build GRCh38
3703657 c.293T>G p.F98C Chr.11: 69055216-69055216 on Build GRCh38
3703656 c.286T>A p.L96M Chr.11: 69055209-69055209 on Build GRCh38
4855761 c.264C>T p.F88F Chr.11: 69055187-69055187 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs562127624 Chr.11: 69055014-69055014 on Build GRCh38
rs376409865 Chr.11: 69055166-69055166 on Build GRCh38
rs185329045 Chr.11: 69055077-69055077 on Build GRCh38
rs759510555 Chr.11: 69055132-69055132 on Build GRCh38
rs866507807 Chr.11: 69055011-69055011 on Build GRCh38
rs566073559 Chr.11: 69055065-69055065 on Build GRCh38
rs980027366 Chr.11: 69055109-69055109 on Build GRCh38
rs180720917 Chr.11: 69054986-69054986 on Build GRCh38
rs116215960 Chr.11: 69055086-69055086 on Build GRCh38
rs926627798 Chr.11: 69054985-69054985 on Build GRCh38
rs529242696 Chr.11: 69055038-69055038 on Build GRCh38
rs991927040 Chr.11: 69055172-69055172 on Build GRCh38
rs1008372314 Chr.11: 69055019-69055019 on Build GRCh38
rs779361967 Chr.11: 69055093-69055093 on Build GRCh38
rs765535152 Chr.11: 69055137-69055137 on Build GRCh38
rs373181469 Chr.11: 69055163-69055163 on Build GRCh38
rs890585457 Chr.11: 69055017-69055017 on Build GRCh38
rs115602952 Chr.11: 69055000-69055000 on Build GRCh38
rs971194862 Chr.11: 69055104-69055104 on Build GRCh38
rs749977548 Chr.11: 69055182-69055182 on Build GRCh38
rs939193188 Chr.11: 69055135-69055135 on Build GRCh38
rs552445433 Chr.11: 69055081-69055081 on Build GRCh38
rs369544894 Chr.11: 69054980-69054980 on Build GRCh38
rs962927046 Chr.11: 69055162-69055162 on Build GRCh38
rs763184008 Chr.11: 69055139-69055139 on Build GRCh38
rs1018724249 Chr.11: 69055020-69055020 on Build GRCh38
rs764375724 Chr.11: 69055147-69055147 on Build GRCh38
rs150386880 Chr.11: 69055184-69055184 on Build GRCh38
rs1009955270 Chr.11: 69055084-69055084 on Build GRCh38
rs1038357785 Chr.11: 69054994-69054994 on Build GRCh38
rs115204200 Chr.11: 69055190-69055190 on Build GRCh38
rs773165448 Chr.11: 69055138-69055138 on Build GRCh38
rs956466723 Chr.11: 69055055-69055055 on Build GRCh38
rs538157837 Chr.11: 69055114-69055114 on Build GRCh38
rs776907811 Chr.11: 69055131-69055131 on Build GRCh38
rs369434788 Chr.11: 69055189-69055189 on Build GRCh38
rs142015442 Chr.11: 69055039-69055039 on Build GRCh38
rs781634412 Chr.11: 69055015-69055015 on Build GRCh38
rs757823111 Chr.11: 69055164-69055164 on Build GRCh38
rs1022414938 Chr.11: 69055076-69055076 on Build GRCh38
rs772129471 Chr.11: 69055127-69055127 on Build GRCh38
rs987878554 Chr.11: 69055056-69055056 on Build GRCh38

View Full Product Details