Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4199892-4200139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD24
Entrez Gene ID
170961
Gene Name
ankyrin repeat domain 24
Gene Aliases
-
UniGene
Hs.197872
Gene Chromosome Location
Chr.19: 4182618-4224814 on Build GRCh38
Transcripts
NM_133475.1 XM_011527756.2 XM_011527757.2 XM_011527758.2 XM_011527761.1 XM_011527764.2 XM_011527765.2 XM_017026438.1 XM_017026439.1 XM_017026440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4661805 c.269C>T p.A90V Chr.19: 4200097-4200097 on Build GRCh38
7355582 c.221T>C p.V74A Chr.19: 4199972-4199972 on Build GRCh38
6585355 c.241G>A p.E81K Chr.19: 4199992-4199992 on Build GRCh38
6585356 c.511G>A p.E171K Chr.19: 4199992-4199992 on Build GRCh38
4661806 c.539C>T p.A180V Chr.19: 4200097-4200097 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535029289 Chr.19: 4199928-4199928 on Build GRCh38
rs867748391 Chr.19: 4199994-4199994 on Build GRCh38
rs751583616 Chr.19: 4200103-4200103 on Build GRCh38
rs768292158 Chr.19: 4200006-4200007 on Build GRCh38
rs765780384 Chr.19: 4200006-4200006 on Build GRCh38
rs200378118 Chr.19: 4200055-4200055 on Build GRCh38
rs748972060 Chr.19: 4200023-4200023 on Build GRCh38
rs370556051 Chr.19: 4200015-4200015 on Build GRCh38
rs762034429 Chr.19: 4200094-4200094 on Build GRCh38
rs373580986 Chr.19: 4199972-4199972 on Build GRCh38
rs745874881 Chr.19: 4199938-4199938 on Build GRCh38
rs896711768 Chr.19: 4200003-4200003 on Build GRCh38
rs750989263 Chr.19: 4200013-4200013 on Build GRCh38
rs571673470 Chr.19: 4200100-4200100 on Build GRCh38
rs571877115 Chr.19: 4199955-4199955 on Build GRCh38
rs865815277 Chr.19: 4199917-4199917 on Build GRCh38
rs776356949 Chr.19: 4200092-4200092 on Build GRCh38
rs188123256 Chr.19: 4200056-4200056 on Build GRCh38
rs900322134 Chr.19: 4200072-4200072 on Build GRCh38
rs370256655 Chr.19: 4200027-4200027 on Build GRCh38
rs61743270 Chr.19: 4199947-4199947 on Build GRCh38
rs768125887 Chr.19: 4200081-4200081 on Build GRCh38
rs369715879 Chr.19: 4199979-4199979 on Build GRCh38
rs371983246 Chr.19: 4199929-4199929 on Build GRCh38
rs781216347 Chr.19: 4200109-4200109 on Build GRCh38
rs537651280 Chr.19: 4199956-4199956 on Build GRCh38
rs773807594 Chr.19: 4200079-4200079 on Build GRCh38
rs368880588 Chr.19: 4199960-4199960 on Build GRCh38
rs995561124 Chr.19: 4200063-4200063 on Build GRCh38
rs199499488 Chr.19: 4200102-4200102 on Build GRCh38
rs770777570 Chr.19: 4200025-4200025 on Build GRCh38
rs374318872 Chr.19: 4200095-4200095 on Build GRCh38
rs373157110 Chr.19: 4199991-4199991 on Build GRCh38
rs374943322 Chr.19: 4200114-4200114 on Build GRCh38
rs758566007 Chr.19: 4199939-4199939 on Build GRCh38
rs375393067 Chr.19: 4199978-4199978 on Build GRCh38
rs371791696 Chr.19: 4200111-4200111 on Build GRCh38
rs924560520 Chr.19: 4199983-4199983 on Build GRCh38
rs555065740 Chr.19: 4200050-4200050 on Build GRCh38
rs111250660 Chr.19: 4200116-4200116 on Build GRCh38
rs377295871 Chr.19: 4200005-4200005 on Build GRCh38
rs1027186138 Chr.19: 4200076-4200076 on Build GRCh38
rs374689174 Chr.19: 4200028-4200028 on Build GRCh38
rs772858858 Chr.19: 4199950-4199950 on Build GRCh38
rs143744058 Chr.19: 4199926-4199926 on Build GRCh38
rs201105748 Chr.19: 4200101-4200101 on Build GRCh38
rs574188339 Chr.19: 4200017-4200017 on Build GRCh38
rs539849986 Chr.19: 4199946-4199946 on Build GRCh38
rs749162502 Chr.19: 4199952-4199952 on Build GRCh38
rs201520760 Chr.19: 4200022-4200022 on Build GRCh38
rs768959278 Chr.19: 4199949-4199949 on Build GRCh38
rs755138595 Chr.19: 4200018-4200018 on Build GRCh38
rs868711993 Chr.19: 4199915-4199915 on Build GRCh38
rs533115261 Chr.19: 4200110-4200110 on Build GRCh38
rs45544832 Chr.19: 4200016-4200016 on Build GRCh38
rs755092650 Chr.19: 4200108-4200108 on Build GRCh38

View Full Product Details