Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3137332-3137567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF213
Entrez Gene ID
7760
Gene Name
zinc finger protein 213
Gene Aliases
CR53, ZKSCAN21, ZSCAN53
UniGene
Hs.115284
Gene Chromosome Location
Chr.16: 3135056-3142804 on Build GRCh38
Transcripts
NM_001134655.1 NM_004220.2 NR_104432.1 XM_011522652.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3509310 c.216C>G p.R72R Chr.16: 3137496-3137496 on Build GRCh38
6567983 c.153C>G p.F51L Chr.16: 3137433-3137433 on Build GRCh38
1731470 c.212_223del12 p.C71_R75>W Chr.16: 3137492-3137503 on Build GRCh38
3509309 c.90G>A p.Q30Q Chr.16: 3137370-3137370 on Build GRCh38
307675 c.83G>T p.W28L Chr.16: 3137363-3137363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962375734 Chr.16: 3137484-3137484 on Build GRCh38
rs141332740 Chr.16: 3137403-3137403 on Build GRCh38
rs748030675 Chr.16: 3137404-3137404 on Build GRCh38
rs746027703 Chr.16: 3137500-3137500 on Build GRCh38
rs756113667 Chr.16: 3137472-3137472 on Build GRCh38
rs764862392 Chr.16: 3137445-3137445 on Build GRCh38
rs749303187 Chr.16: 3137494-3137494 on Build GRCh38
rs893831056 Chr.16: 3137414-3137414 on Build GRCh38
rs970091140 Chr.16: 3137465-3137465 on Build GRCh38
rs933770671 Chr.16: 3137506-3137506 on Build GRCh38
rs774160035 Chr.16: 3137539-3137539 on Build GRCh38
rs918302108 Chr.16: 3137496-3137496 on Build GRCh38
rs775984835 Chr.16: 3137443-3137443 on Build GRCh38
rs959536471 Chr.16: 3137434-3137434 on Build GRCh38
rs527638676 Chr.16: 3137361-3137361 on Build GRCh38
rs1026903193 Chr.16: 3137451-3137451 on Build GRCh38
rs766035827 Chr.16: 3137450-3137451 on Build GRCh38
rs761115835 Chr.16: 3137444-3137444 on Build GRCh38
rs750107521 Chr.16: 3137384-3137384 on Build GRCh38
rs781076086 Chr.16: 3137462-3137462 on Build GRCh38
rs943600549 Chr.16: 3137542-3137542 on Build GRCh38
rs143487323 Chr.16: 3137371-3137371 on Build GRCh38
rs751175576 Chr.16: 3137459-3137459 on Build GRCh38
rs747584057 Chr.16: 3137509-3137509 on Build GRCh38
rs7197270 Chr.16: 3137430-3137430 on Build GRCh38
rs537954118 Chr.16: 3137450-3137450 on Build GRCh38
rs764664544 Chr.16: 3137379-3137379 on Build GRCh38
rs202036376 Chr.16: 3137458-3137458 on Build GRCh38
rs201883457 Chr.16: 3137425-3137425 on Build GRCh38
rs200158720 Chr.16: 3137426-3137426 on Build GRCh38
rs777946424 Chr.16: 3137478-3137478 on Build GRCh38
rs35685765 Chr.16: 3137542-3137543 on Build GRCh38
rs772829298 Chr.16: 3137511-3137511 on Build GRCh38
rs762708025 Chr.16: 3137457-3137457 on Build GRCh38
rs151095978 Chr.16: 3137382-3137382 on Build GRCh38
rs770006514 Chr.16: 3137413-3137413 on Build GRCh38
rs754825183 Chr.16: 3137460-3137460 on Build GRCh38
rs779968309 Chr.16: 3137394-3137394 on Build GRCh38
rs370856670 Chr.16: 3137420-3137420 on Build GRCh38
rs757975776 Chr.16: 3137393-3137393 on Build GRCh38
rs1051928429 Chr.16: 3137519-3137519 on Build GRCh38
rs751006475 Chr.16: 3137494-3137495 on Build GRCh38
rs202142918 Chr.16: 3137471-3137471 on Build GRCh38
rs762393927 Chr.16: 3137516-3137516 on Build GRCh38
rs770990282 Chr.16: 3137495-3137495 on Build GRCh38
rs775821363 Chr.16: 3137504-3137504 on Build GRCh38
rs909807098 Chr.16: 3137481-3137481 on Build GRCh38
rs566807578 Chr.16: 3137419-3137419 on Build GRCh38
rs755016107 Chr.16: 3137398-3137398 on Build GRCh38
rs765873120 Chr.16: 3137535-3137535 on Build GRCh38
rs531525413 Chr.16: 3137397-3137397 on Build GRCh38
rs779031082 Chr.16: 3137499-3137499 on Build GRCh38
rs568922381 Chr.16: 3137439-3137439 on Build GRCh38
rs772442197 Chr.16: 3137503-3137503 on Build GRCh38

View Full Product Details