Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 145052439-145052710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf33
Entrez Gene ID
65265
Gene Name
chromosome 8 open reading frame 33
Gene Aliases
-
UniGene
Hs.169615
Gene Chromosome Location
Chr.8: 145052438-145056030 on Build GRCh38
Transcripts
NM_023080.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5430943 c.53G>T p.G18V Chr.8: 145052632-145052632 on Build GRCh38
6616691 c.87C>A p.P29P Chr.8: 145052666-145052666 on Build GRCh38
5625704 c.51G>T p.P17P Chr.8: 145052630-145052630 on Build GRCh38
4162871 c.124T>C p.C42R Chr.8: 145052703-145052703 on Build GRCh38
1097358 c.72C>T p.R24R Chr.8: 145052651-145052651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768405966 Chr.8: 145052467-145052467 on Build GRCh38
rs772307682 Chr.8: 145052685-145052685 on Build GRCh38
rs201947442 Chr.8: 145052687-145052687 on Build GRCh38
rs763250391 Chr.8: 145052461-145052462 on Build GRCh38
rs757833232 Chr.8: 145052570-145052571 on Build GRCh38
rs756603502 Chr.8: 145052569-145052569 on Build GRCh38
rs761674574 Chr.8: 145052475-145052475 on Build GRCh38
rs576048175 Chr.8: 145052615-145052615 on Build GRCh38
rs758154681 Chr.8: 145052657-145052657 on Build GRCh38
rs777694600 Chr.8: 145052587-145052587 on Build GRCh38
rs772057544 Chr.8: 145052586-145052586 on Build GRCh38
rs371372179 Chr.8: 145052666-145052666 on Build GRCh38
rs369663195 Chr.8: 145052555-145052555 on Build GRCh38
rs759735303 Chr.8: 145052477-145052477 on Build GRCh38
rs773320342 Chr.8: 145052533-145052533 on Build GRCh38
rs773601864 Chr.8: 145052644-145052644 on Build GRCh38
rs375967102 Chr.8: 145052463-145052463 on Build GRCh38
rs761178929 Chr.8: 145052604-145052604 on Build GRCh38
rs537396574 Chr.8: 145052600-145052600 on Build GRCh38
rs772217850 Chr.8: 145052639-145052639 on Build GRCh38
rs771965907 Chr.8: 145052528-145052528 on Build GRCh38
rs771015168 Chr.8: 145052623-145052623 on Build GRCh38
rs1051188363 Chr.8: 145052473-145052473 on Build GRCh38
rs767499143 Chr.8: 145052476-145052476 on Build GRCh38
rs372741321 Chr.8: 145052535-145052535 on Build GRCh38
rs760982820 Chr.8: 145052647-145052647 on Build GRCh38
rs142863079 Chr.8: 145052682-145052682 on Build GRCh38
rs751234000 Chr.8: 145052661-145052661 on Build GRCh38
rs200194569 Chr.8: 145052534-145052534 on Build GRCh38
rs199517196 Chr.8: 145052617-145052617 on Build GRCh38
rs764336430 Chr.8: 145052542-145052542 on Build GRCh38
rs777714721 Chr.8: 145052618-145052618 on Build GRCh38
rs763039582 Chr.8: 145052596-145052596 on Build GRCh38
rs774283607 Chr.8: 145052635-145052635 on Build GRCh38
rs755662728 Chr.8: 145052517-145052517 on Build GRCh38
rs551205680 Chr.8: 145052519-145052519 on Build GRCh38
rs1047050064 Chr.8: 145052499-145052499 on Build GRCh38
rs149359042 Chr.8: 145052652-145052652 on Build GRCh38
rs139779460 Chr.8: 145052614-145052614 on Build GRCh38
rs775462688 Chr.8: 145052590-145052590 on Build GRCh38
rs768849032 Chr.8: 145052629-145052629 on Build GRCh38
rs774676416 Chr.8: 145052598-145052598 on Build GRCh38
rs139941405 Chr.8: 145052671-145052671 on Build GRCh38
rs767838512 Chr.8: 145052550-145052550 on Build GRCh38
rs559199391 Chr.8: 145052527-145052527 on Build GRCh38
rs765374384 Chr.8: 145052478-145052478 on Build GRCh38
rs752926746 Chr.8: 145052481-145052481 on Build GRCh38
rs762215591 Chr.8: 145052547-145052547 on Build GRCh38
rs201350395 Chr.8: 145052575-145052575 on Build GRCh38
rs201179746 Chr.8: 145052466-145052466 on Build GRCh38
rs199825158 Chr.8: 145052656-145052656 on Build GRCh38
rs768598047 Chr.8: 145052597-145052597 on Build GRCh38
rs572434877 Chr.8: 145052489-145052489 on Build GRCh38
rs144574967 Chr.8: 145052650-145052650 on Build GRCh38
rs769459679 Chr.8: 145052465-145052465 on Build GRCh38
rs748190702 Chr.8: 145052683-145052683 on Build GRCh38
rs199714906 Chr.8: 145052574-145052574 on Build GRCh38
rs763089817 Chr.8: 145052539-145052539 on Build GRCh38
rs775580418 Chr.8: 145052537-145052537 on Build GRCh38
rs781628211 Chr.8: 145052508-145052508 on Build GRCh38
rs764569578 Chr.8: 145052509-145052511 on Build GRCh38
rs763789746 Chr.8: 145052658-145052658 on Build GRCh38
rs746955353 Chr.8: 145052588-145052588 on Build GRCh38
rs751892687 Chr.8: 145052546-145052546 on Build GRCh38
rs868411265 Chr.8: 145052668-145052668 on Build GRCh38
rs373931353 Chr.8: 145052514-145052514 on Build GRCh38
rs144602242 Chr.8: 145052610-145052610 on Build GRCh38
rs1001048733 Chr.8: 145052554-145052554 on Build GRCh38
rs758727551 Chr.8: 145052458-145052458 on Build GRCh38
rs566306900 Chr.8: 145052523-145052523 on Build GRCh38
rs1039221487 Chr.8: 145052510-145052510 on Build GRCh38
rs755950327 Chr.8: 145052672-145052672 on Build GRCh38
rs542954845 Chr.8: 145052464-145052464 on Build GRCh38
rs551375087 Chr.8: 145052624-145052624 on Build GRCh38
rs754553736 Chr.8: 145052612-145052612 on Build GRCh38
rs765503458 Chr.8: 145052457-145052457 on Build GRCh38
rs1004920487 Chr.8: 145052643-145052643 on Build GRCh38
rs542655933 Chr.8: 145052571-145052571 on Build GRCh38
rs748953887 Chr.8: 145052520-145052520 on Build GRCh38
rs781239458 Chr.8: 145052627-145052627 on Build GRCh38
rs757736460 Chr.8: 145052498-145052498 on Build GRCh38
rs377526568 Chr.8: 145052526-145052526 on Build GRCh38
rs900029259 Chr.8: 145052552-145052552 on Build GRCh38
rs747526913 Chr.8: 145052462-145052462 on Build GRCh38
rs777257800 Chr.8: 145052651-145052651 on Build GRCh38
rs751883651 Chr.8: 145052493-145052493 on Build GRCh38
rs376982061 Chr.8: 145052619-145052619 on Build GRCh38
rs751843050 Chr.8: 145052543-145052543 on Build GRCh38
rs778193901 Chr.8: 145052461-145052461 on Build GRCh38
rs149304579 Chr.8: 145052589-145052589 on Build GRCh38
rs533045040 Chr.8: 145052474-145052474 on Build GRCh38
rs764377921 Chr.8: 145052490-145052490 on Build GRCh38
rs371401733 Chr.8: 145052460-145052460 on Build GRCh38
rs767930297 Chr.8: 145052602-145052602 on Build GRCh38

View Full Product Details