Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 11061325-11061580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRH1
Entrez Gene ID
5554
Gene Name
proline rich protein HaeIII subfamily 1
Gene Aliases
Db-s, PA, PIF-S, PRH2, Pr1/Pr2
UniGene
Hs.656965
Gene Chromosome Location
Chr.12: 10880961-11171623 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_133575.1 NR_037918.2 NM_001316893.1 NM_176887.2

Gene Symbol
PRH1-PRR4
Entrez Gene ID
100533464
Gene Name
PRH1-PRR4 readthrough
Gene Aliases
PRH1
UniGene
Hs.732031
Gene Chromosome Location
Chr.12: 10845849-11171625 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_133575.1 NR_037918.2 NM_001316893.1 NM_176887.2

Gene Symbol
PRH1-TAS2R14
Entrez Gene ID
106707243
Gene Name
PRH1-TAS2R14 readthrough
Gene Aliases
-
UniGene
Hs.731575
Gene Chromosome Location
Chr.12: 10937405-11171623 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_133575.1 NR_037918.2 NM_001316893.1 NM_176887.2

Gene Symbol
TAS2R46
Entrez Gene ID
259292
Gene Name
taste 2 receptor member 46
Gene Aliases
T2R46, T2R54
UniGene
Hs.688193
Gene Chromosome Location
Chr.12: 11061365-11062294 on Build GRCh38
Transcripts
NM_001291314.1 NM_001291315.1 NR_133575.1 NR_037918.2 NM_001316893.1 NM_176887.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4594641 c.828A>G p.P276P Chr.12: 11061467-11061467 on Build GRCh38
4602839 c.888T>A p.H296Q Chr.12: 11061407-11061407 on Build GRCh38
6352601 c.834C>G p.I278M Chr.12: 11061461-11061461 on Build GRCh38
6054628 c.893G>C p.R298T Chr.12: 11061402-11061402 on Build GRCh38
4595137 c.761G>A p.S254N Chr.12: 11061534-11061534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553647348 Chr.12: 11061410-11061410 on Build GRCh38
rs764685134 Chr.12: 11061452-11061452 on Build GRCh38
rs76020872 Chr.12: 11061534-11061534 on Build GRCh38
rs777519851 Chr.12: 11061413-11061413 on Build GRCh38
rs557044065 Chr.12: 11061398-11061398 on Build GRCh38
rs768843374 Chr.12: 11061502-11061502 on Build GRCh38
rs775964550 Chr.12: 11061443-11061443 on Build GRCh38
rs76147157 Chr.12: 11061538-11061538 on Build GRCh38
rs757249666 Chr.12: 11061488-11061488 on Build GRCh38
rs370408044 Chr.12: 11061493-11061493 on Build GRCh38
rs941700841 Chr.12: 11061494-11061494 on Build GRCh38
rs749575510 Chr.12: 11061510-11061510 on Build GRCh38
rs746887895 Chr.12: 11061483-11061483 on Build GRCh38
rs376068759 Chr.12: 11061545-11061545 on Build GRCh38
rs372278369 Chr.12: 11061426-11061426 on Build GRCh38
rs769907597 Chr.12: 11061440-11061440 on Build GRCh38
rs561938546 Chr.12: 11061503-11061503 on Build GRCh38
rs762373246 Chr.12: 11061509-11061509 on Build GRCh38
rs771070419 Chr.12: 11061421-11061421 on Build GRCh38
rs754611567 Chr.12: 11061395-11061395 on Build GRCh38
rs754699113 Chr.12: 11061467-11061467 on Build GRCh38
rs553357638 Chr.12: 11061474-11061474 on Build GRCh38
rs764041640 Chr.12: 11061550-11061550 on Build GRCh38
rs763206168 Chr.12: 11061449-11061449 on Build GRCh38
rs150894148 Chr.12: 11061433-11061433 on Build GRCh38
rs747861564 Chr.12: 11061406-11061406 on Build GRCh38
rs764769965 Chr.12: 11061470-11061470 on Build GRCh38
rs753858957 Chr.12: 11061392-11061405 on Build GRCh38
rs749695359 Chr.12: 11061499-11061499 on Build GRCh38
rs755574779 Chr.12: 11061361-11061361 on Build GRCh38
rs772092030 Chr.12: 11061407-11061407 on Build GRCh38
rs747114723 Chr.12: 11061418-11061418 on Build GRCh38
rs753429374 Chr.12: 11061394-11061394 on Build GRCh38
rs553708509 Chr.12: 11061464-11061464 on Build GRCh38
rs200528237 Chr.12: 11061475-11061475 on Build GRCh38
rs774802527 Chr.12: 11061453-11061453 on Build GRCh38
rs776162761 Chr.12: 11061524-11061524 on Build GRCh38
rs777855752 Chr.12: 11061481-11061481 on Build GRCh38
rs757540705 Chr.12: 11061399-11061400 on Build GRCh38
rs773393543 Chr.12: 11061516-11061516 on Build GRCh38
rs777881796 Chr.12: 11061411-11061411 on Build GRCh38
rs2708381 Chr.12: 11061546-11061546 on Build GRCh38
rs796631017 Chr.12: 11061402-11061402 on Build GRCh38
rs758867798 Chr.12: 11061484-11061485 on Build GRCh38
rs2599402 Chr.12: 11061370-11061370 on Build GRCh38
rs185280425 Chr.12: 11061381-11061381 on Build GRCh38
rs781493121 Chr.12: 11061492-11061492 on Build GRCh38
rs750628525 Chr.12: 11061444-11061445 on Build GRCh38
rs368038360 Chr.12: 11061420-11061420 on Build GRCh38
rs778984342 Chr.12: 11061421-11061421 on Build GRCh38
rs752221868 Chr.12: 11061542-11061542 on Build GRCh38
rs778835386 Chr.12: 11061401-11061401 on Build GRCh38
rs760189646 Chr.12: 11061457-11061457 on Build GRCh38
rs139412224 Chr.12: 11061461-11061461 on Build GRCh38
rs977446658 Chr.12: 11061468-11061468 on Build GRCh38
rs542162895 Chr.12: 11061496-11061496 on Build GRCh38

View Full Product Details