Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20974422-20974671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIFM3
Entrez Gene ID
150209
Gene Name
apoptosis inducing factor, mitochondria associated 3
Gene Aliases
AIFL
UniGene
Hs.723246
Gene Chromosome Location
Chr.22: 20965130-20981360 on Build GRCh38
Transcripts
NM_001018060.2 NM_001146288.1 NM_144704.2 NR_027464.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3552363 c.571+2T>C p.? Chr.22: 20974623-20974623 on Build GRCh38
3552361 c.503T>C p.V168A Chr.22: 20974553-20974553 on Build GRCh38
4439253 c.547A>C p.K183Q Chr.22: 20974561-20974561 on Build GRCh38
4411181 c.519G>T p.Q173H Chr.22: 20974533-20974533 on Build GRCh38
4439255 c.511A>C p.K171Q Chr.22: 20974561-20974561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151146728 Chr.22: 20974621-20974621 on Build GRCh38
rs752514354 Chr.22: 20974563-20974563 on Build GRCh38
rs777066175 Chr.22: 20974479-20974479 on Build GRCh38
rs144287643 Chr.22: 20974549-20974549 on Build GRCh38
rs780326587 Chr.22: 20974628-20974628 on Build GRCh38
rs144284647 Chr.22: 20974610-20974610 on Build GRCh38
rs545189474 Chr.22: 20974449-20974449 on Build GRCh38
rs188620860 Chr.22: 20974453-20974453 on Build GRCh38
rs368597263 Chr.22: 20974481-20974481 on Build GRCh38
rs1002942426 Chr.22: 20974472-20974472 on Build GRCh38
rs758018566 Chr.22: 20974565-20974565 on Build GRCh38
rs538743781 Chr.22: 20974636-20974636 on Build GRCh38
rs142117053 Chr.22: 20974540-20974540 on Build GRCh38
rs770126928 Chr.22: 20974495-20974495 on Build GRCh38
rs200165374 Chr.22: 20974614-20974614 on Build GRCh38
rs878866227 Chr.22: 20974445-20974445 on Build GRCh38
rs373567342 Chr.22: 20974499-20974499 on Build GRCh38
rs201015871 Chr.22: 20974587-20974587 on Build GRCh38
rs761930853 Chr.22: 20974527-20974527 on Build GRCh38
rs373611938 Chr.22: 20974649-20974649 on Build GRCh38
rs764350593 Chr.22: 20974521-20974521 on Build GRCh38
rs770315903 Chr.22: 20974626-20974626 on Build GRCh38
rs777356244 Chr.22: 20974569-20974569 on Build GRCh38
rs766430143 Chr.22: 20974532-20974532 on Build GRCh38
rs568287583 Chr.22: 20974469-20974469 on Build GRCh38
rs1021391494 Chr.22: 20974648-20974648 on Build GRCh38
rs759511916 Chr.22: 20974541-20974541 on Build GRCh38
rs774600076 Chr.22: 20974526-20974526 on Build GRCh38
rs749621144 Chr.22: 20974633-20974633 on Build GRCh38

View Full Product Details