Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96839656-96839923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD23
Entrez Gene ID
200539
Gene Name
ankyrin repeat domain 23
Gene Aliases
DARP, MARP3
UniGene
Hs.643430
Gene Chromosome Location
Chr.2: 96837914-96844021 on Build GRCh38
Transcripts
NM_144994.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1238186 c.783A>G p.L261L Chr.2: 96839766-96839766 on Build GRCh38
2825849 c.815G>A p.R272Q Chr.2: 96839734-96839734 on Build GRCh38
722817 c.801C>T p.A267A Chr.2: 96839748-96839748 on Build GRCh38
1410101 c.802G>A p.E268K Chr.2: 96839747-96839747 on Build GRCh38
3991527 c.727G>A p.G243R Chr.2: 96839822-96839822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755698341 Chr.2: 96839798-96839798 on Build GRCh38
rs776561347 Chr.2: 96839822-96839822 on Build GRCh38
rs564570522 Chr.2: 96839817-96839817 on Build GRCh38
rs771080613 Chr.2: 96839747-96839747 on Build GRCh38
rs752413804 Chr.2: 96839765-96839765 on Build GRCh38
rs938836635 Chr.2: 96839748-96839748 on Build GRCh38
rs575883381 Chr.2: 96839712-96839712 on Build GRCh38
rs554184995 Chr.2: 96839825-96839825 on Build GRCh38
rs780672957 Chr.2: 96839794-96839794 on Build GRCh38
rs775729933 Chr.2: 96839837-96839837 on Build GRCh38
rs938164758 Chr.2: 96839709-96839709 on Build GRCh38
rs772606680 Chr.2: 96839689-96839689 on Build GRCh38
rs748927330 Chr.2: 96839800-96839800 on Build GRCh38
rs769105015 Chr.2: 96839718-96839719 on Build GRCh38
rs374745685 Chr.2: 96839693-96839693 on Build GRCh38
rs151227731 Chr.2: 96839785-96839785 on Build GRCh38
rs762310851 Chr.2: 96839792-96839799 on Build GRCh38
rs765717120 Chr.2: 96839808-96839809 on Build GRCh38
rs776622207 Chr.2: 96839790-96839790 on Build GRCh38
rs901092174 Chr.2: 96839904-96839904 on Build GRCh38
rs145736274 Chr.2: 96839774-96839774 on Build GRCh38
rs766472569 Chr.2: 96839872-96839873 on Build GRCh38
rs1017424558 Chr.2: 96839856-96839856 on Build GRCh38
rs149329617 Chr.2: 96839730-96839730 on Build GRCh38
rs779455378 Chr.2: 96839871-96839871 on Build GRCh38
rs533182462 Chr.2: 96839860-96839860 on Build GRCh38
rs939700678 Chr.2: 96839849-96839849 on Build GRCh38
rs368094328 Chr.2: 96839838-96839838 on Build GRCh38
rs758257387 Chr.2: 96839706-96839706 on Build GRCh38
rs745443017 Chr.2: 96839727-96839727 on Build GRCh38
rs372499217 Chr.2: 96839715-96839715 on Build GRCh38
rs541144314 Chr.2: 96839789-96839789 on Build GRCh38
rs1056376690 Chr.2: 96839753-96839753 on Build GRCh38
rs985974299 Chr.2: 96839855-96839855 on Build GRCh38
rs372229183 Chr.2: 96839899-96839899 on Build GRCh38
rs763809902 Chr.2: 96839840-96839840 on Build GRCh38
rs914567696 Chr.2: 96839696-96839696 on Build GRCh38
rs755612425 Chr.2: 96839863-96839863 on Build GRCh38
rs780034472 Chr.2: 96839713-96839713 on Build GRCh38
rs771634323 Chr.2: 96839819-96839819 on Build GRCh38
rs200423018 Chr.2: 96839773-96839773 on Build GRCh38
rs369796952 Chr.2: 96839861-96839861 on Build GRCh38
rs780693506 Chr.2: 96839719-96839719 on Build GRCh38
rs140909554 Chr.2: 96839740-96839740 on Build GRCh38
rs777116196 Chr.2: 96839781-96839782 on Build GRCh38
rs370902350 Chr.2: 96839842-96839842 on Build GRCh38
rs756707778 Chr.2: 96839717-96839717 on Build GRCh38
rs766600523 Chr.2: 96839690-96839690 on Build GRCh38
rs752606484 Chr.2: 96839701-96839701 on Build GRCh38
rs769794770 Chr.2: 96839835-96839835 on Build GRCh38
rs748592857 Chr.2: 96839729-96839729 on Build GRCh38
rs928982719 Chr.2: 96839870-96839872 on Build GRCh38
rs763087506 Chr.2: 96839869-96839869 on Build GRCh38
rs539381940 Chr.2: 96839675-96839675 on Build GRCh38
rs201589615 Chr.2: 96839694-96839694 on Build GRCh38
rs984972873 Chr.2: 96839903-96839903 on Build GRCh38
rs776856475 Chr.2: 96839752-96839752 on Build GRCh38
rs779792830 Chr.2: 96839799-96839799 on Build GRCh38
rs977685877 Chr.2: 96839724-96839724 on Build GRCh38
rs759673941 Chr.2: 96839755-96839755 on Build GRCh38
rs949871540 Chr.2: 96839789-96839803 on Build GRCh38
rs761290975 Chr.2: 96839742-96839742 on Build GRCh38
rs772518278 Chr.2: 96839809-96839809 on Build GRCh38
rs965951842 Chr.2: 96839856-96839856 on Build GRCh38
rs140411651 Chr.2: 96839796-96839796 on Build GRCh38
rs779130904 Chr.2: 96839728-96839728 on Build GRCh38
rs374551685 Chr.2: 96839811-96839811 on Build GRCh38
rs765555617 Chr.2: 96839760-96839760 on Build GRCh38

View Full Product Details