Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 28086111-28086370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF165
Entrez Gene ID
7718
Gene Name
zinc finger protein 165
Gene Aliases
CT53, LD65, ZSCAN7
UniGene
Hs.55481
Gene Chromosome Location
Chr.6: 28080704-28104244 on Build GRCh38
Transcripts
NM_003447.3 XM_017011258.1 XM_017011259.1 XM_017011260.1 XM_017011261.1 XM_017011262.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5869944 c.454G>A p.V152M Chr.6: 28086214-28086214 on Build GRCh38
483786 c.490C>G p.Q164E Chr.6: 28086250-28086250 on Build GRCh38
165750 c.424G>A p.E142K Chr.6: 28086184-28086184 on Build GRCh38
741664 c.412-1G>A p.? Chr.6: 28086171-28086171 on Build GRCh38
6828172 c.548G>C p.C183S Chr.6: 28086308-28086308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200718368 Chr.6: 28086200-28086200 on Build GRCh38
rs573432778 Chr.6: 28086227-28086227 on Build GRCh38
rs760183176 Chr.6: 28086261-28086261 on Build GRCh38
rs747134688 Chr.6: 28086152-28086152 on Build GRCh38
rs759521504 Chr.6: 28086303-28086303 on Build GRCh38
rs752557753 Chr.6: 28086312-28086312 on Build GRCh38
rs765148128 Chr.6: 28086305-28086305 on Build GRCh38
rs770479587 Chr.6: 28086343-28086343 on Build GRCh38
rs774862941 Chr.6: 28086345-28086346 on Build GRCh38
rs148774905 Chr.6: 28086224-28086224 on Build GRCh38
rs913506029 Chr.6: 28086148-28086148 on Build GRCh38
rs750767704 Chr.6: 28086169-28086169 on Build GRCh38
rs778052971 Chr.6: 28086146-28086146 on Build GRCh38
rs753318770 Chr.6: 28086281-28086281 on Build GRCh38
rs955508581 Chr.6: 28086170-28086170 on Build GRCh38
rs558228867 Chr.6: 28086340-28086340 on Build GRCh38
rs781371300 Chr.6: 28086338-28086338 on Build GRCh38
rs746407502 Chr.6: 28086178-28086178 on Build GRCh38
rs765949235 Chr.6: 28086280-28086280 on Build GRCh38
rs987008730 Chr.6: 28086175-28086175 on Build GRCh38
rs759453234 Chr.6: 28086282-28086282 on Build GRCh38
rs202004589 Chr.6: 28086198-28086198 on Build GRCh38
rs781083050 Chr.6: 28086162-28086162 on Build GRCh38
rs771055826 Chr.6: 28086157-28086157 on Build GRCh38
rs376282649 Chr.6: 28086347-28086347 on Build GRCh38
rs780700024 Chr.6: 28086346-28086346 on Build GRCh38
rs1028330186 Chr.6: 28086221-28086221 on Build GRCh38
rs772918276 Chr.6: 28086240-28086240 on Build GRCh38
rs957350487 Chr.6: 28086187-28086187 on Build GRCh38
rs771686454 Chr.6: 28086239-28086239 on Build GRCh38
rs763287452 Chr.6: 28086196-28086196 on Build GRCh38
rs955791770 Chr.6: 28086294-28086294 on Build GRCh38
rs757380347 Chr.6: 28086334-28086334 on Build GRCh38
rs751828438 Chr.6: 28086331-28086331 on Build GRCh38
rs553657777 Chr.6: 28086214-28086214 on Build GRCh38
rs758195240 Chr.6: 28086328-28086328 on Build GRCh38
rs538645613 Chr.6: 28086330-28086330 on Build GRCh38

View Full Product Details