Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 82317271-82317542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD7
Entrez Gene ID
924
Gene Name
CD7 molecule
Gene Aliases
GP40, LEU-9, TP41, Tp40
UniGene
Hs.36972
Gene Chromosome Location
Chr.17: 82314865-82320829 on Build GRCh38
Transcripts
NM_006137.6 XM_011523608.1 XM_017025316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4638724 c.55G>T p.G19C Chr.17: 82317441-82317441 on Build GRCh38
6426097 c.20T>C p.L7P Chr.17: 82317476-82317476 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779686442 Chr.17: 82317379-82317379 on Build GRCh38
rs3176829 Chr.17: 82317377-82317377 on Build GRCh38
rs760031043 Chr.17: 82317452-82317452 on Build GRCh38
rs771956071 Chr.17: 82317442-82317442 on Build GRCh38
rs371661907 Chr.17: 82317297-82317297 on Build GRCh38
rs749101827 Chr.17: 82317398-82317398 on Build GRCh38
rs929976901 Chr.17: 82317513-82317513 on Build GRCh38
rs528736203 Chr.17: 82317378-82317378 on Build GRCh38
rs760225055 Chr.17: 82317481-82317481 on Build GRCh38
rs547000288 Chr.17: 82317391-82317391 on Build GRCh38
rs916648099 Chr.17: 82317463-82317463 on Build GRCh38
rs773521834 Chr.17: 82317367-82317367 on Build GRCh38
rs372973906 Chr.17: 82317441-82317441 on Build GRCh38
rs1008766667 Chr.17: 82317296-82317296 on Build GRCh38
rs1044255902 Chr.17: 82317462-82317462 on Build GRCh38
rs766003973 Chr.17: 82317482-82317482 on Build GRCh38
rs770256884 Chr.17: 82317461-82317461 on Build GRCh38
rs749904144 Chr.17: 82317479-82317479 on Build GRCh38
rs773179275 Chr.17: 82317443-82317443 on Build GRCh38
rs941486738 Chr.17: 82317471-82317471 on Build GRCh38
rs767170031 Chr.17: 82317376-82317376 on Build GRCh38
rs778710030 Chr.17: 82317500-82317500 on Build GRCh38
rs575431051 Chr.17: 82317312-82317312 on Build GRCh38
rs374780838 Chr.17: 82317451-82317451 on Build GRCh38
rs753237279 Chr.17: 82317373-82317373 on Build GRCh38
rs771126775 Chr.17: 82317449-82317449 on Build GRCh38
rs989562058 Chr.17: 82317438-82317439 on Build GRCh38
rs775961541 Chr.17: 82317464-82317464 on Build GRCh38
rs964534389 Chr.17: 82317330-82317330 on Build GRCh38
rs752433050 Chr.17: 82317522-82317522 on Build GRCh38
rs747044187 Chr.17: 82317444-82317444 on Build GRCh38
rs542852261 Chr.17: 82317348-82317348 on Build GRCh38
rs897117424 Chr.17: 82317496-82317496 on Build GRCh38
rs764600064 Chr.17: 82317477-82317477 on Build GRCh38
rs765906706 Chr.17: 82317372-82317372 on Build GRCh38
rs763573567 Chr.17: 82317469-82317469 on Build GRCh38
rs532176048 Chr.17: 82317470-82317470 on Build GRCh38
rs974915781 Chr.17: 82317302-82317302 on Build GRCh38
rs754807783 Chr.17: 82317489-82317489 on Build GRCh38
rs754752718 Chr.17: 82317399-82317399 on Build GRCh38
rs756869795 Chr.17: 82317374-82317374 on Build GRCh38
rs867697412 Chr.17: 82317491-82317491 on Build GRCh38
rs765529795 Chr.17: 82317490-82317490 on Build GRCh38
rs759975995 Chr.17: 82317371-82317371 on Build GRCh38
rs760830225 Chr.17: 82317368-82317368 on Build GRCh38
rs778869218 Chr.17: 82317414-82317414 on Build GRCh38
rs146365930 Chr.17: 82317487-82317487 on Build GRCh38

View Full Product Details