Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179379208-179379473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AXDND1
Entrez Gene ID
126859
Gene Name
axonemal dynein light chain domain containing 1
Gene Aliases
C1orf125
UniGene
Hs.658505
Gene Chromosome Location
Chr.1: 179365705-179554735 on Build GRCh38
Transcripts
NM_144696.5 NR_073544.1 XM_011509165.1 XM_011509166.2 XM_011509167.2 XM_011509168.1 XM_011509169.1 XM_011509170.1 XM_011509171.1 XM_011509174.1 XM_011509175.1 XM_011509176.1 XM_011509178.2 XM_011509179.2 XM_011509180.1 XM_017000257.1 XM_017000258.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
900254 c.565T>G p.L189V Chr.1: 179379466-179379466 on Build GRCh38
2095309 c.501G>T p.K167N Chr.1: 179379402-179379402 on Build GRCh38
5825810 c.501G>T p.K167N Chr.1: 179379402-179379402 on Build GRCh38
1689303 c.535C>T p.H179Y Chr.1: 179379436-179379436 on Build GRCh38
4788594 c.549T>C p.S183S Chr.1: 179379450-179379450 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538753394 Chr.1: 179379315-179379315 on Build GRCh38
rs1003586275 Chr.1: 179379422-179379422 on Build GRCh38
rs368898032 Chr.1: 179379353-179379353 on Build GRCh38
rs1034981398 Chr.1: 179379439-179379439 on Build GRCh38
rs771701974 Chr.1: 179379440-179379446 on Build GRCh38
rs918821730 Chr.1: 179379279-179379279 on Build GRCh38
rs773447411 Chr.1: 179379235-179379235 on Build GRCh38
rs1010465018 Chr.1: 179379371-179379371 on Build GRCh38
rs747758324 Chr.1: 179379359-179379359 on Build GRCh38
rs193279432 Chr.1: 179379400-179379400 on Build GRCh38
rs756374321 Chr.1: 179379351-179379351 on Build GRCh38
rs1041687806 Chr.1: 179379399-179379399 on Build GRCh38
rs555912852 Chr.1: 179379281-179379281 on Build GRCh38
rs972986550 Chr.1: 179379260-179379260 on Build GRCh38
rs777614715 Chr.1: 179379365-179379365 on Build GRCh38
rs188751199 Chr.1: 179379320-179379320 on Build GRCh38
rs536540513 Chr.1: 179379248-179379248 on Build GRCh38
rs1050714506 Chr.1: 179379309-179379309 on Build GRCh38
rs995391213 Chr.1: 179379340-179379340 on Build GRCh38
rs764119003 Chr.1: 179379388-179379388 on Build GRCh38
rs769430363 Chr.1: 179379435-179379435 on Build GRCh38
rs368998259 Chr.1: 179379403-179379403 on Build GRCh38
rs754407481 Chr.1: 179379405-179379405 on Build GRCh38
rs373376663 Chr.1: 179379361-179379361 on Build GRCh38
rs775386430 Chr.1: 179379436-179379436 on Build GRCh38
rs569604544 Chr.1: 179379308-179379308 on Build GRCh38
rs761159257 Chr.1: 179379249-179379249 on Build GRCh38
rs746501655 Chr.1: 179379368-179379368 on Build GRCh38
rs780197866 Chr.1: 179379379-179379383 on Build GRCh38
rs372275076 Chr.1: 179379334-179379334 on Build GRCh38

View Full Product Details