Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 89114193-89114428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIGD2
Entrez Gene ID
166815
Gene Name
tigger transposable element derived 2
Gene Aliases
HEL106
UniGene
Hs.58924
Gene Chromosome Location
Chr.4: 89112817-89114901 on Build GRCh38
Transcripts
NM_145715.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7318980 c.1297G>C p.D433H Chr.4: 89114271-89114271 on Build GRCh38
6047264 c.1263A>G p.E421E Chr.4: 89114237-89114237 on Build GRCh38
420478 c.1304G>T p.R435L Chr.4: 89114278-89114278 on Build GRCh38
3428766 c.1375C>T p.Q459* Chr.4: 89114349-89114349 on Build GRCh38
7393843 c.1273A>G p.I425V Chr.4: 89114247-89114247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761439839 Chr.4: 89114283-89114283 on Build GRCh38
rs778581378 Chr.4: 89114367-89114367 on Build GRCh38
rs763383072 Chr.4: 89114336-89114336 on Build GRCh38
rs935797382 Chr.4: 89114301-89114301 on Build GRCh38
rs751801454 Chr.4: 89114346-89114346 on Build GRCh38
rs865923569 Chr.4: 89114337-89114337 on Build GRCh38
rs771647880 Chr.4: 89114379-89114379 on Build GRCh38
rs757425154 Chr.4: 89114349-89114349 on Build GRCh38
rs1023436538 Chr.4: 89114382-89114382 on Build GRCh38
rs765873866 Chr.4: 89114324-89114324 on Build GRCh38
rs769734342 Chr.4: 89114270-89114270 on Build GRCh38
rs779775119 Chr.4: 89114271-89114271 on Build GRCh38
rs772613693 Chr.4: 89114316-89114316 on Build GRCh38
rs768437030 Chr.4: 89114365-89114365 on Build GRCh38
rs765641733 Chr.4: 89114234-89114234 on Build GRCh38
rs753138691 Chr.4: 89114236-89114236 on Build GRCh38
rs138689511 Chr.4: 89114357-89114357 on Build GRCh38
rs2280099 Chr.4: 89114398-89114398 on Build GRCh38
rs757343884 Chr.4: 89114248-89114248 on Build GRCh38
rs750489906 Chr.4: 89114351-89114351 on Build GRCh38
rs539533845 Chr.4: 89114278-89114278 on Build GRCh38
rs747783057 Chr.4: 89114369-89114369 on Build GRCh38
rs758651005 Chr.4: 89114239-89114239 on Build GRCh38
rs760074283 Chr.4: 89114317-89114317 on Build GRCh38
rs771548167 Chr.4: 89114296-89114296 on Build GRCh38
rs530878158 Chr.4: 89114371-89114371 on Build GRCh38
rs144834721 Chr.4: 89114247-89114247 on Build GRCh38
rs764607775 Chr.4: 89114343-89114343 on Build GRCh38
rs776123234 Chr.4: 89114403-89114403 on Build GRCh38
rs367929262 Chr.4: 89114256-89114256 on Build GRCh38
rs779712032 Chr.4: 89114386-89114386 on Build GRCh38
rs141253259 Chr.4: 89114307-89114307 on Build GRCh38
rs749281435 Chr.4: 89114359-89114359 on Build GRCh38
rs146343241 Chr.4: 89114258-89114258 on Build GRCh38
rs757441263 Chr.4: 89114276-89114277 on Build GRCh38
rs1000129223 Chr.4: 89114390-89114390 on Build GRCh38
rs753294270 Chr.4: 89114334-89114334 on Build GRCh38
rs375311520 Chr.4: 89114319-89114319 on Build GRCh38
rs756227518 Chr.4: 89114355-89114355 on Build GRCh38
rs530462147 Chr.4: 89114225-89114225 on Build GRCh38
rs765359788 Chr.4: 89114284-89114285 on Build GRCh38
rs200957545 Chr.4: 89114277-89114277 on Build GRCh38
rs922535241 Chr.4: 89114263-89114263 on Build GRCh38
rs1041574360 Chr.4: 89114275-89114275 on Build GRCh38

View Full Product Details