Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 24256544-24256799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG3
Entrez Gene ID
10368
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 3
Gene Aliases
-
UniGene
Hs.7235
Gene Chromosome Location
Chr.16: 24255553-24362416 on Build GRCh38
Transcripts
NM_006539.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5674202 c.19G>C p.G7R Chr.16: 24256773-24256773 on Build GRCh38
3361697 c.20G>A p.G7D Chr.16: 24256774-24256774 on Build GRCh38
4435409 c.17G>A p.R6K Chr.16: 24256771-24256771 on Build GRCh38
7426766 c.38C>A p.T13N Chr.16: 24256792-24256792 on Build GRCh38
319197 c.5G>C p.R2T Chr.16: 24256759-24256759 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750619795 Chr.16: 24256755-24256757 on Build GRCh38
rs765885399 Chr.16: 24256706-24256706 on Build GRCh38
rs368874081 Chr.16: 24256708-24256708 on Build GRCh38
rs368009855 Chr.16: 24256645-24256645 on Build GRCh38
rs938194480 Chr.16: 24256602-24256602 on Build GRCh38
rs762662582 Chr.16: 24256564-24256564 on Build GRCh38
rs554899263 Chr.16: 24256581-24256581 on Build GRCh38
rs755987658 Chr.16: 24256716-24256716 on Build GRCh38
rs371728578 Chr.16: 24256712-24256712 on Build GRCh38
rs542929447 Chr.16: 24256660-24256660 on Build GRCh38
rs901066179 Chr.16: 24256743-24256743 on Build GRCh38
rs766905446 Chr.16: 24256751-24256751 on Build GRCh38
rs928198502 Chr.16: 24256585-24256585 on Build GRCh38
rs775615485 Chr.16: 24256736-24256736 on Build GRCh38
rs1056338928 Chr.16: 24256678-24256678 on Build GRCh38
rs1048546449 Chr.16: 24256696-24256696 on Build GRCh38
rs558770318 Chr.16: 24256668-24256668 on Build GRCh38
rs567328927 Chr.16: 24256655-24256655 on Build GRCh38
rs778953751 Chr.16: 24256717-24256717 on Build GRCh38
rs990953303 Chr.16: 24256608-24256608 on Build GRCh38
rs892295887 Chr.16: 24256689-24256689 on Build GRCh38
rs773865647 Chr.16: 24256705-24256705 on Build GRCh38
rs754955162 Chr.16: 24256710-24256710 on Build GRCh38
rs867380526 Chr.16: 24256723-24256723 on Build GRCh38
rs945093162 Chr.16: 24256695-24256695 on Build GRCh38
rs537810531 Chr.16: 24256732-24256732 on Build GRCh38
rs757811826 Chr.16: 24256704-24256704 on Build GRCh38
rs931052437 Chr.16: 24256670-24256670 on Build GRCh38
rs772222270 Chr.16: 24256733-24256733 on Build GRCh38
rs909186417 Chr.16: 24256568-24256568 on Build GRCh38
rs746132287 Chr.16: 24256721-24256721 on Build GRCh38

View Full Product Details