Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2484435-2484694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF343
Entrez Gene ID
79175
Gene Name
zinc finger protein 343
Gene Aliases
dJ734P14.5
UniGene
Hs.516846
Gene Chromosome Location
Chr.20: 2481817-2524522 on Build GRCh38
Transcripts
NM_001282495.1 NM_001282496.1 NM_001282497.1 NM_001282498.1 NM_001321800.1 NM_001321801.1 NM_001321802.1 NM_001321803.1 NM_001321805.1 NM_024325.5 XM_017028062.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5563324 c.469C>T p.Q157* Chr.20: 2484492-2484492 on Build GRCh38
2887203 c.388C>G p.Q130E Chr.20: 2484573-2484573 on Build GRCh38
1410829 c.486C>T p.S162S Chr.20: 2484475-2484475 on Build GRCh38
1410830 c.456A>G p.K152K Chr.20: 2484505-2484505 on Build GRCh38
7248336 c.448C>T p.H150Y Chr.20: 2484513-2484513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542750943 Chr.20: 2484471-2484471 on Build GRCh38
rs150915057 Chr.20: 2484567-2484567 on Build GRCh38
rs762796135 Chr.20: 2484586-2484586 on Build GRCh38
rs1047197016 Chr.20: 2484543-2484543 on Build GRCh38
rs139390703 Chr.20: 2484574-2484574 on Build GRCh38
rs113667110 Chr.20: 2484576-2484576 on Build GRCh38
rs919560452 Chr.20: 2484458-2484458 on Build GRCh38
rs773144691 Chr.20: 2484615-2484615 on Build GRCh38
rs147664383 Chr.20: 2484570-2484570 on Build GRCh38
rs200150089 Chr.20: 2484659-2484659 on Build GRCh38
rs766666312 Chr.20: 2484634-2484634 on Build GRCh38
rs756279815 Chr.20: 2484518-2484518 on Build GRCh38
rs746015433 Chr.20: 2484618-2484618 on Build GRCh38
rs535922881 Chr.20: 2484563-2484563 on Build GRCh38
rs755423125 Chr.20: 2484557-2484557 on Build GRCh38
rs754222312 Chr.20: 2484555-2484555 on Build GRCh38
rs73085335 Chr.20: 2484658-2484658 on Build GRCh38
rs201212283 Chr.20: 2484500-2484500 on Build GRCh38
rs775432868 Chr.20: 2484508-2484508 on Build GRCh38
rs147660623 Chr.20: 2484655-2484655 on Build GRCh38
rs764152510 Chr.20: 2484609-2484609 on Build GRCh38
rs770545655 Chr.20: 2484573-2484573 on Build GRCh38
rs539865734 Chr.20: 2484581-2484581 on Build GRCh38
rs779513588 Chr.20: 2484560-2484560 on Build GRCh38
rs749569536 Chr.20: 2484585-2484586 on Build GRCh38
rs568275863 Chr.20: 2484509-2484509 on Build GRCh38
rs534495790 Chr.20: 2484479-2484479 on Build GRCh38
rs886839790 Chr.20: 2484564-2484564 on Build GRCh38
rs750583060 Chr.20: 2484516-2484516 on Build GRCh38
rs766758120 Chr.20: 2484519-2484519 on Build GRCh38
rs769593787 Chr.20: 2484580-2484580 on Build GRCh38

View Full Product Details