Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 103075334-103075595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC10
Entrez Gene ID
83787
Gene Name
armadillo repeat containing 10
Gene Aliases
PNAS-112, PNAS112, PSEC0198, SVH
UniGene
Hs.287412
Gene Chromosome Location
Chr.7: 103074727-103099763 on Build GRCh38
Transcripts
NM_001161009.2 NM_001161010.2 NM_001161011.2 NM_001161012.2 NM_001161013.2 NM_031905.4 XM_005250628.2 XM_011516601.2 XM_017012684.1 XM_017012685.1 XM_017012686.1 XM_017012688.1 XM_017012689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3749993 c.85C>T p.R29W Chr.7: 103075357-103075357 on Build GRCh38
6401638 c.121C>T p.R41C Chr.7: 103075393-103075393 on Build GRCh38
395360 c.96_97GC>TT p.R32>? Chr.7: 103075368-103075369 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779255956 Chr.7: 103075401-103075401 on Build GRCh38
rs371290359 Chr.7: 103075462-103075462 on Build GRCh38
rs894199784 Chr.7: 103075482-103075482 on Build GRCh38
rs902022616 Chr.7: 103075546-103075546 on Build GRCh38
rs1055881945 Chr.7: 103075472-103075472 on Build GRCh38
rs766952064 Chr.7: 103075456-103075456 on Build GRCh38
rs776469655 Chr.7: 103075358-103075358 on Build GRCh38
rs867572603 Chr.7: 103075540-103075540 on Build GRCh38
rs999057977 Chr.7: 103075536-103075536 on Build GRCh38
rs770589668 Chr.7: 103075451-103075451 on Build GRCh38
rs566426253 Chr.7: 103075436-103075436 on Build GRCh38
rs1054710572 Chr.7: 103075470-103075470 on Build GRCh38
rs892080857 Chr.7: 103075486-103075486 on Build GRCh38
rs747076972 Chr.7: 103075422-103075422 on Build GRCh38
rs548740984 Chr.7: 103075450-103075450 on Build GRCh38
rs975417834 Chr.7: 103075568-103075568 on Build GRCh38
rs1011764408 Chr.7: 103075511-103075511 on Build GRCh38
rs537569156 Chr.7: 103075459-103075459 on Build GRCh38
rs762729207 Chr.7: 103075366-103075366 on Build GRCh38
rs769767254 Chr.7: 103075362-103075362 on Build GRCh38
rs746871233 Chr.7: 103075356-103075356 on Build GRCh38
rs749092208 Chr.7: 103075429-103075429 on Build GRCh38
rs539699561 Chr.7: 103075566-103075566 on Build GRCh38
rs889445719 Chr.7: 103075570-103075570 on Build GRCh38
rs533777688 Chr.7: 103075438-103075438 on Build GRCh38
rs201282030 Chr.7: 103075393-103075393 on Build GRCh38
rs567336124 Chr.7: 103075455-103075455 on Build GRCh38
rs967550619 Chr.7: 103075530-103075530 on Build GRCh38
rs775642271 Chr.7: 103075415-103075415 on Build GRCh38
rs765760923 Chr.7: 103075370-103075370 on Build GRCh38
rs768631257 Chr.7: 103075441-103075441 on Build GRCh38
rs368052104 Chr.7: 103075421-103075421 on Build GRCh38
rs781260563 Chr.7: 103075432-103075432 on Build GRCh38
rs946217349 Chr.7: 103075503-103075503 on Build GRCh38
rs1021735264 Chr.7: 103075522-103075523 on Build GRCh38
rs764659282 Chr.7: 103075389-103075389 on Build GRCh38
rs778652302 Chr.7: 103075453-103075454 on Build GRCh38
rs966021080 Chr.7: 103075553-103075553 on Build GRCh38
rs909840272 Chr.7: 103075355-103075355 on Build GRCh38
rs757561720 Chr.7: 103075400-103075400 on Build GRCh38
rs7798381 Chr.7: 103075357-103075357 on Build GRCh38
rs768681814 Chr.7: 103075423-103075423 on Build GRCh38
rs750602776 Chr.7: 103075408-103075408 on Build GRCh38
rs182065746 Chr.7: 103075510-103075510 on Build GRCh38
rs746408605 Chr.7: 103075461-103075461 on Build GRCh38
rs557497443 Chr.7: 103075489-103075489 on Build GRCh38
rs1002509516 Chr.7: 103075467-103075467 on Build GRCh38
rs773166453 Chr.7: 103075444-103075444 on Build GRCh38
rs780188553 Chr.7: 103075418-103075418 on Build GRCh38
rs761691628 Chr.7: 103075359-103075359 on Build GRCh38
rs1048870564 Chr.7: 103075427-103075427 on Build GRCh38
rs975820716 Chr.7: 103075407-103075407 on Build GRCh38
rs780087962 Chr.7: 103075395-103075395 on Build GRCh38
rs751063064 Chr.7: 103075386-103075386 on Build GRCh38
rs759011482 Chr.7: 103075387-103075387 on Build GRCh38
rs1019783252 Chr.7: 103075547-103075547 on Build GRCh38
rs998306649 Chr.7: 103075559-103075559 on Build GRCh38
rs777057694 Chr.7: 103075457-103075457 on Build GRCh38

View Full Product Details