Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6530806-6531067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFSF9
Entrez Gene ID
8744
Gene Name
tumor necrosis factor superfamily member 9
Gene Aliases
4-1BB-L, CD137L, TNLG5A
UniGene
Hs.1524
Gene Chromosome Location
Chr.19: 6530999-6535928 on Build GRCh38
Transcripts
NM_003811.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4581548 c.23C>T p.S8L Chr.19: 6531059-6531059 on Build GRCh38
139113 c.3G>A p.M1I Chr.19: 6531039-6531039 on Build GRCh38
3404748 c.18C>T p.D6D Chr.19: 6531054-6531054 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs561643259 Chr.19: 6531021-6531021 on Build GRCh38
rs964069287 Chr.19: 6530955-6530955 on Build GRCh38
rs1025859202 Chr.19: 6530934-6530934 on Build GRCh38
rs577004619 Chr.19: 6530961-6530961 on Build GRCh38
rs554769790 Chr.19: 6530900-6530900 on Build GRCh38
rs1050968318 Chr.19: 6530831-6530831 on Build GRCh38
rs200266919 Chr.19: 6531023-6531023 on Build GRCh38
rs752838037 Chr.19: 6530898-6530898 on Build GRCh38
rs771972170 Chr.19: 6531026-6531026 on Build GRCh38
rs774185243 Chr.19: 6531022-6531022 on Build GRCh38
rs780614625 Chr.19: 6531010-6531010 on Build GRCh38
rs1001753441 Chr.19: 6530891-6530891 on Build GRCh38
rs775299991 Chr.19: 6531029-6531029 on Build GRCh38
rs1039636869 Chr.19: 6531004-6531004 on Build GRCh38
rs775168247 Chr.19: 6531013-6531014 on Build GRCh38
rs1023913305 Chr.19: 6531045-6531045 on Build GRCh38
rs778965612 Chr.19: 6530858-6530858 on Build GRCh38
rs746348686 Chr.19: 6530996-6530996 on Build GRCh38
rs761263674 Chr.19: 6531040-6531040 on Build GRCh38
rs568176970 Chr.19: 6530909-6530909 on Build GRCh38
rs760453634 Chr.19: 6531031-6531031 on Build GRCh38
rs896671551 Chr.19: 6531034-6531034 on Build GRCh38
rs763754219 Chr.19: 6531033-6531033 on Build GRCh38
rs546156218 Chr.19: 6530924-6530943 on Build GRCh38
rs370221849 Chr.19: 6531015-6531015 on Build GRCh38
rs758605424 Chr.19: 6530937-6530937 on Build GRCh38
rs995301577 Chr.19: 6530903-6530903 on Build GRCh38
rs769066726 Chr.19: 6531014-6531014 on Build GRCh38
rs553922465 Chr.19: 6530946-6530946 on Build GRCh38
rs1027764685 Chr.19: 6530929-6530929 on Build GRCh38
rs779295452 Chr.19: 6530993-6530993 on Build GRCh38
rs748189289 Chr.19: 6531019-6531019 on Build GRCh38
rs935159260 Chr.19: 6530976-6530976 on Build GRCh38
rs546126926 Chr.19: 6530971-6530971 on Build GRCh38
rs529771596 Chr.19: 6530867-6530867 on Build GRCh38
rs753467032 Chr.19: 6531035-6531035 on Build GRCh38
rs749879984 Chr.19: 6531047-6531047 on Build GRCh38
rs891076985 Chr.19: 6530836-6530836 on Build GRCh38
rs991106985 Chr.19: 6530957-6530957 on Build GRCh38
rs1012903087 Chr.19: 6531043-6531043 on Build GRCh38
rs2234173 Chr.19: 6531017-6531017 on Build GRCh38
rs933947434 Chr.19: 6530969-6530969 on Build GRCh38
rs764756760 Chr.19: 6531046-6531046 on Build GRCh38
rs753351795 Chr.19: 6530988-6530988 on Build GRCh38
rs758857666 Chr.19: 6531006-6531006 on Build GRCh38
rs946098834 Chr.19: 6530978-6530978 on Build GRCh38
rs2234172 Chr.19: 6530979-6530979 on Build GRCh38
rs541767564 Chr.19: 6531011-6531011 on Build GRCh38
rs1014821756 Chr.19: 6530922-6530922 on Build GRCh38
rs1006768383 Chr.19: 6530837-6530837 on Build GRCh38
rs1025784050 Chr.19: 6531024-6531024 on Build GRCh38
rs61760043 Chr.19: 6530986-6530986 on Build GRCh38
rs1005092409 Chr.19: 6530938-6530938 on Build GRCh38
rs913769025 Chr.19: 6530975-6530975 on Build GRCh38
rs975326523 Chr.19: 6530960-6530960 on Build GRCh38
rs756736578 Chr.19: 6530992-6530992 on Build GRCh38

View Full Product Details