Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64078936-64079163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRGAP1
Entrez Gene ID
57522
Gene Name
SLIT-ROBO Rho GTPase activating protein 1
Gene Aliases
ARHGAP13, NMTC2
UniGene
Hs.210751
Gene Chromosome Location
Chr.12: 63844372-64147863 on Build GRCh38
Transcripts
NM_020762.2 XM_005269042.3 XM_011538580.1 XM_011538581.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6073164 c.1209G>T p.Q403H Chr.12: 64079002-64079002 on Build GRCh38
1363575 c.1224A>G p.T408T Chr.12: 64079017-64079017 on Build GRCh38
3735819 c.1321_1323delATG p.M441delM Chr.12: 64079114-64079116 on Build GRCh38
6588161 c.1225G>A p.E409K Chr.12: 64079018-64079018 on Build GRCh38
1990949 c.1157C>T p.T386M Chr.12: 64078950-64078950 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764609174 Chr.12: 64078980-64078980 on Build GRCh38
rs767296904 Chr.12: 64079125-64079125 on Build GRCh38
rs753813324 Chr.12: 64079134-64079134 on Build GRCh38
rs374046573 Chr.12: 64079134-64079135 on Build GRCh38
rs747123507 Chr.12: 64079104-64079104 on Build GRCh38
rs1014529237 Chr.12: 64079138-64079138 on Build GRCh38
rs777173697 Chr.12: 64079109-64079109 on Build GRCh38
rs765629579 Chr.12: 64079121-64079121 on Build GRCh38
rs762283825 Chr.12: 64079110-64079110 on Build GRCh38
rs756063357 Chr.12: 64079032-64079032 on Build GRCh38
rs757065762 Chr.12: 64079051-64079051 on Build GRCh38
rs752266023 Chr.12: 64079031-64079031 on Build GRCh38
rs789710 Chr.12: 64079023-64079023 on Build GRCh38
rs754113179 Chr.12: 64078998-64078998 on Build GRCh38
rs549661090 Chr.12: 64079044-64079044 on Build GRCh38
rs773451694 Chr.12: 64079122-64079122 on Build GRCh38
rs752210935 Chr.12: 64079127-64079127 on Build GRCh38
rs376912099 Chr.12: 64079096-64079096 on Build GRCh38
rs955486119 Chr.12: 64079095-64079095 on Build GRCh38
rs267603622 Chr.12: 64078991-64078991 on Build GRCh38
rs762207000 Chr.12: 64079000-64079000 on Build GRCh38
rs778642960 Chr.12: 64079053-64079053 on Build GRCh38
rs767735923 Chr.12: 64079103-64079104 on Build GRCh38
rs763574419 Chr.12: 64079131-64079131 on Build GRCh38
rs746037667 Chr.12: 64079068-64079068 on Build GRCh38
rs759204851 Chr.12: 64079124-64079124 on Build GRCh38
rs777794012 Chr.12: 64079033-64079033 on Build GRCh38
rs775500354 Chr.12: 64079074-64079074 on Build GRCh38
rs1043493648 Chr.12: 64079024-64079024 on Build GRCh38
rs755650058 Chr.12: 64079128-64079128 on Build GRCh38
rs765568604 Chr.12: 64079009-64079009 on Build GRCh38
rs751210214 Chr.12: 64079010-64079010 on Build GRCh38
rs761113797 Chr.12: 64078972-64078972 on Build GRCh38
rs780779940 Chr.12: 64079029-64079029 on Build GRCh38
rs768762999 Chr.12: 64079106-64079106 on Build GRCh38
rs114669147 Chr.12: 64079073-64079073 on Build GRCh38

View Full Product Details