Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73500752-73500977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP3CB-AS1
Entrez Gene ID
101929145
Gene Name
PPP3CB antisense RNA 1 (head to head)
Gene Aliases
USP54-AS1
UniGene
Hs.434502
Gene Chromosome Location
Chr.10: 73495752-73507309 on Build GRCh38
Transcripts
NR_132103.1 NR_132104.1 NM_001320437.1 NM_152586.3 NR_135249.1 NR_135250.1 XM_005269582.2 XM_011539368.2 XM_017015759.1 XM_017015760.1 XM_017015761.1 XM_017015762.1 XM_017015763.1 XM_017015764.1 XM_017015765.1 XM_017015766.1 XM_017015767.1 XM_017015768.1 XM_017015769.1 XM_017015770.1 XM_017015771.1 XM_017015772.1 XM_017015773.1 XM_017015774.1 XM_017015775.1 XM_017015776.1 XM_017015777.1 XM_017015778.1 XM_017015779.1 XM_017015780.1 XM_017015781.1 XM_017015782.1 XM_017015783.1 XM_017015784.1 XM_017015785.1 XM_017015786.1

Gene Symbol
USP54
Entrez Gene ID
159195
Gene Name
ubiquitin specific peptidase 54
Gene Aliases
C10orf29, bA137L10.3, bA137L10.4
UniGene
Hs.657355
Gene Chromosome Location
Chr.10: 73497538-73626844 on Build GRCh38
Transcripts
NR_132103.1 NR_132104.1 NM_001320437.1 NM_152586.3 NR_135249.1 NR_135250.1 XM_005269582.2 XM_011539368.2 XM_017015759.1 XM_017015760.1 XM_017015761.1 XM_017015762.1 XM_017015763.1 XM_017015764.1 XM_017015765.1 XM_017015766.1 XM_017015767.1 XM_017015768.1 XM_017015769.1 XM_017015770.1 XM_017015771.1 XM_017015772.1 XM_017015773.1 XM_017015774.1 XM_017015775.1 XM_017015776.1 XM_017015777.1 XM_017015778.1 XM_017015779.1 XM_017015780.1 XM_017015781.1 XM_017015782.1 XM_017015783.1 XM_017015784.1 XM_017015785.1 XM_017015786.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4780353 c.4327A>C p.R1443R Chr.10: 73500823-73500823 on Build GRCh38
6483736 c.4345G>A p.G1449R Chr.10: 73500805-73500805 on Build GRCh38
1745319 c.1593_1595delCGG p.G532delG Chr.10: 73500804-73500806 on Build GRCh38
7325002 c.4376C>A p.P1459H Chr.10: 73500774-73500774 on Build GRCh38
7325001 c.1625C>A p.P542H Chr.10: 73500774-73500774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748102722 Chr.10: 73500812-73500812 on Build GRCh38
rs150856920 Chr.10: 73500827-73500827 on Build GRCh38
rs374734222 Chr.10: 73500780-73500780 on Build GRCh38
rs746930517 Chr.10: 73500835-73500835 on Build GRCh38
rs1037855917 Chr.10: 73500808-73500808 on Build GRCh38
rs751312300 Chr.10: 73500800-73500800 on Build GRCh38
rs777066643 Chr.10: 73500866-73500866 on Build GRCh38
rs111294104 Chr.10: 73500805-73500805 on Build GRCh38
rs530927755 Chr.10: 73500921-73500921 on Build GRCh38
rs138141992 Chr.10: 73500799-73500799 on Build GRCh38
rs750114103 Chr.10: 73500783-73500783 on Build GRCh38
rs920384502 Chr.10: 73500817-73500817 on Build GRCh38
rs1029437511 Chr.10: 73500918-73500918 on Build GRCh38
rs770456951 Chr.10: 73500865-73500866 on Build GRCh38
rs758004808 Chr.10: 73500798-73500798 on Build GRCh38
rs762381005 Chr.10: 73500870-73500870 on Build GRCh38
rs141723921 Chr.10: 73500806-73500806 on Build GRCh38
rs367720957 Chr.10: 73500852-73500852 on Build GRCh38
rs890677589 Chr.10: 73500895-73500895 on Build GRCh38
rs1050067988 Chr.10: 73500884-73500884 on Build GRCh38
rs764661905 Chr.10: 73500881-73500881 on Build GRCh38
rs1041827442 Chr.10: 73500922-73500922 on Build GRCh38
rs1015891512 Chr.10: 73500902-73500902 on Build GRCh38
rs547431371 Chr.10: 73500937-73500937 on Build GRCh38
rs184258285 Chr.10: 73500826-73500826 on Build GRCh38
rs772374256 Chr.10: 73500832-73500832 on Build GRCh38
rs201611515 Chr.10: 73500856-73500856 on Build GRCh38
rs146601363 Chr.10: 73500903-73500903 on Build GRCh38
rs976967503 Chr.10: 73500910-73500910 on Build GRCh38
rs112673115 Chr.10: 73500944-73500944 on Build GRCh38

View Full Product Details