Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36480564-36480829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TXN2
Entrez Gene ID
25828
Gene Name
thioredoxin 2
Gene Aliases
COXPD29, MT-TRX, MTRX, TRX2
UniGene
Hs.211929
Gene Chromosome Location
Chr.22: 36467036-36482025 on Build GRCh38
Transcripts
NM_012473.3 XM_005261508.1 XM_006724226.1 XM_011530111.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5579918 c.119G>A p.G40D Chr.22: 36480719-36480719 on Build GRCh38
5393536 c.92C>T p.A31V Chr.22: 36480746-36480746 on Build GRCh38
5424046 c.189T>G p.N63K Chr.22: 36480649-36480649 on Build GRCh38
1308131 c.61C>G p.Q21E Chr.22: 36480777-36480777 on Build GRCh38
6234249 c.179C>A p.T60K Chr.22: 36480659-36480659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772829928 Chr.22: 36480621-36480621 on Build GRCh38
rs1053223731 Chr.22: 36480756-36480756 on Build GRCh38
rs776346675 Chr.22: 36480730-36480730 on Build GRCh38
rs1023266570 Chr.22: 36480800-36480800 on Build GRCh38
rs11545777 Chr.22: 36480746-36480746 on Build GRCh38
rs780135728 Chr.22: 36480759-36480759 on Build GRCh38
rs748133874 Chr.22: 36480709-36480709 on Build GRCh38
rs758960286 Chr.22: 36480732-36480732 on Build GRCh38
rs774277776 Chr.22: 36480601-36480601 on Build GRCh38
rs904578603 Chr.22: 36480699-36480699 on Build GRCh38
rs900861220 Chr.22: 36480747-36480747 on Build GRCh38
rs765889740 Chr.22: 36480719-36480719 on Build GRCh38
rs772949536 Chr.22: 36480716-36480716 on Build GRCh38
rs754527088 Chr.22: 36480671-36480671 on Build GRCh38
rs142583599 Chr.22: 36480752-36480752 on Build GRCh38
rs760541625 Chr.22: 36480717-36480717 on Build GRCh38
rs762374008 Chr.22: 36480742-36480742 on Build GRCh38
rs377748933 Chr.22: 36480646-36480646 on Build GRCh38
rs761591917 Chr.22: 36480602-36480602 on Build GRCh38
rs61730811 Chr.22: 36480673-36480673 on Build GRCh38
rs759410715 Chr.22: 36480790-36480790 on Build GRCh38
rs945943168 Chr.22: 36480616-36480616 on Build GRCh38
rs754147721 Chr.22: 36480760-36480760 on Build GRCh38
rs951207146 Chr.22: 36480783-36480783 on Build GRCh38
rs199512391 Chr.22: 36480751-36480751 on Build GRCh38
rs764895866 Chr.22: 36480734-36480734 on Build GRCh38
rs145571413 Chr.22: 36480688-36480688 on Build GRCh38
rs757680572 Chr.22: 36480676-36480676 on Build GRCh38
rs199746093 Chr.22: 36480733-36480733 on Build GRCh38
rs774261417 Chr.22: 36480708-36480708 on Build GRCh38
rs1043255460 Chr.22: 36480695-36480695 on Build GRCh38
rs777701106 Chr.22: 36480788-36480788 on Build GRCh38
rs780339679 Chr.22: 36480689-36480689 on Build GRCh38
rs368576238 Chr.22: 36480773-36480773 on Build GRCh38
rs371207531 Chr.22: 36480674-36480674 on Build GRCh38
rs891869180 Chr.22: 36480594-36480594 on Build GRCh38
rs770822455 Chr.22: 36480799-36480799 on Build GRCh38
rs895797773 Chr.22: 36480758-36480758 on Build GRCh38
rs749260640 Chr.22: 36480690-36480690 on Build GRCh38
rs369451382 Chr.22: 36480599-36480599 on Build GRCh38
rs374527574 Chr.22: 36480639-36480639 on Build GRCh38
rs754022333 Chr.22: 36480767-36480767 on Build GRCh38
rs766084648 Chr.22: 36480651-36480651 on Build GRCh38
rs61730810 Chr.22: 36480805-36480805 on Build GRCh38
rs781720947 Chr.22: 36480681-36480681 on Build GRCh38
rs61730813 Chr.22: 36480670-36480670 on Build GRCh38
rs11545778 Chr.22: 36480736-36480736 on Build GRCh38
rs146269725 Chr.22: 36480682-36480682 on Build GRCh38
rs368246545 Chr.22: 36480712-36480712 on Build GRCh38
rs148453808 Chr.22: 36480702-36480702 on Build GRCh38
rs767359634 Chr.22: 36480620-36480620 on Build GRCh38
rs771945917 Chr.22: 36480779-36480779 on Build GRCh38
rs534944987 Chr.22: 36480803-36480803 on Build GRCh38
rs746713860 Chr.22: 36480795-36480795 on Build GRCh38
rs372840739 Chr.22: 36480735-36480735 on Build GRCh38

View Full Product Details