Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2539619-2539852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRAT1
Entrez Gene ID
221927
Gene Name
BRCA1 associated ATM activator 1
Gene Aliases
BAAT1, C7orf27, RMFSL
UniGene
Hs.520623
Gene Chromosome Location
Chr.7: 2537810-2555758 on Build GRCh38
Transcripts
NM_152743.3 XM_005249643.4 XM_011515177.2 XM_011515178.1 XM_011515179.2 XM_011515181.2 XM_011515182.2 XM_011515183.1 XM_011515184.2 XM_011515186.2 XM_017011833.1 XM_017011834.1 XM_017011835.1 XM_017011836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4922922 c.1439G>A p.S480N Chr.7: 2539845-2539845 on Build GRCh38
3366898 c.1507C>T p.P503S Chr.7: 2539634-2539634 on Build GRCh38
5997334 c.1480A>G p.I494V Chr.7: 2539804-2539804 on Build GRCh38
7314285 c.1498+10C>A p.? Chr.7: 2539776-2539776 on Build GRCh38
6377137 c.1471G>A p.G491S Chr.7: 2539813-2539813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746362295 Chr.7: 2539785-2539785 on Build GRCh38
rs775762160 Chr.7: 2539688-2539688 on Build GRCh38
rs753735820 Chr.7: 2539758-2539758 on Build GRCh38
rs936655077 Chr.7: 2539769-2539769 on Build GRCh38
rs370879134 Chr.7: 2539650-2539650 on Build GRCh38
rs1013936313 Chr.7: 2539747-2539747 on Build GRCh38
rs746669517 Chr.7: 2539781-2539781 on Build GRCh38
rs765157563 Chr.7: 2539776-2539776 on Build GRCh38
rs774605061 Chr.7: 2539745-2539745 on Build GRCh38
rs758085579 Chr.7: 2539662-2539662 on Build GRCh38
rs767624248 Chr.7: 2539757-2539757 on Build GRCh38
rs777070793 Chr.7: 2539679-2539679 on Build GRCh38
rs749698079 Chr.7: 2539799-2539799 on Build GRCh38
rs145727035 Chr.7: 2539817-2539817 on Build GRCh38
rs770155018 Chr.7: 2539686-2539686 on Build GRCh38
rs142667367 Chr.7: 2539797-2539797 on Build GRCh38
rs752652217 Chr.7: 2539821-2539821 on Build GRCh38
rs371715895 Chr.7: 2539759-2539759 on Build GRCh38
rs555986902 Chr.7: 2539829-2539829 on Build GRCh38
rs1038985112 Chr.7: 2539711-2539711 on Build GRCh38
rs375052234 Chr.7: 2539740-2539740 on Build GRCh38
rs906327946 Chr.7: 2539823-2539823 on Build GRCh38
rs752491146 Chr.7: 2539779-2539779 on Build GRCh38
rs777447384 Chr.7: 2539664-2539664 on Build GRCh38
rs780693251 Chr.7: 2539782-2539782 on Build GRCh38
rs773303692 Chr.7: 2539813-2539813 on Build GRCh38
rs761869109 Chr.7: 2539746-2539746 on Build GRCh38
rs762040027 Chr.7: 2539812-2539812 on Build GRCh38
rs577945739 Chr.7: 2539800-2539800 on Build GRCh38
rs774567258 Chr.7: 2539767-2539767 on Build GRCh38
rs199506042 Chr.7: 2539676-2539676 on Build GRCh38
rs200248064 Chr.7: 2539833-2539833 on Build GRCh38
rs940883846 Chr.7: 2539670-2539670 on Build GRCh38
rs913159579 Chr.7: 2539824-2539824 on Build GRCh38
rs539902160 Chr.7: 2539804-2539804 on Build GRCh38
rs113400040 Chr.7: 2539681-2539681 on Build GRCh38
rs368323897 Chr.7: 2539741-2539741 on Build GRCh38
rs972272299 Chr.7: 2539702-2539702 on Build GRCh38
rs758246723 Chr.7: 2539780-2539780 on Build GRCh38
rs770243304 Chr.7: 2539794-2539794 on Build GRCh38
rs150268284 Chr.7: 2539806-2539806 on Build GRCh38
rs778795414 Chr.7: 2539649-2539649 on Build GRCh38
rs763936681 Chr.7: 2539828-2539828 on Build GRCh38
rs762907691 Chr.7: 2539826-2539826 on Build GRCh38
rs538096996 Chr.7: 2539726-2539726 on Build GRCh38
rs898740738 Chr.7: 2539724-2539724 on Build GRCh38
rs770442549 Chr.7: 2539674-2539674 on Build GRCh38

View Full Product Details