Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40853005-40853258 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPINT1
Entrez Gene ID
6692
Gene Name
serine peptidase inhibitor, Kunitz type 1
Gene Aliases
HAI, HAI1, MANSC2
UniGene
Hs.233950
Gene Chromosome Location
Chr.15: 40844018-40857655 on Build GRCh38
Transcripts
NM_001032367.1 NM_003710.3 NM_181642.2 XM_006720657.1 XM_011521957.1 XM_011521958.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7245004 c.476-6C>T p.? Chr.15: 40853118-40853118 on Build GRCh38
5891497 c.533C>T p.P178L Chr.15: 40853181-40853181 on Build GRCh38
6789016 c.540G>A p.V180V Chr.15: 40853188-40853188 on Build GRCh38
3815988 c.529G>A p.E177K Chr.15: 40853177-40853177 on Build GRCh38
4186037 c.587C>T p.T196M Chr.15: 40853235-40853235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750341676 Chr.15: 40853223-40853223 on Build GRCh38
rs928034005 Chr.15: 40853069-40853069 on Build GRCh38
rs145330357 Chr.15: 40853236-40853236 on Build GRCh38
rs376378235 Chr.15: 40853115-40853115 on Build GRCh38
rs763597848 Chr.15: 40853194-40853194 on Build GRCh38
rs978576098 Chr.15: 40853157-40853157 on Build GRCh38
rs775321975 Chr.15: 40853184-40853184 on Build GRCh38
rs371185922 Chr.15: 40853152-40853152 on Build GRCh38
rs747228024 Chr.15: 40853147-40853147 on Build GRCh38
rs755237849 Chr.15: 40853235-40853235 on Build GRCh38
rs978314111 Chr.15: 40853058-40853058 on Build GRCh38
rs911588797 Chr.15: 40853034-40853034 on Build GRCh38
rs755628866 Chr.15: 40853087-40853087 on Build GRCh38
rs373996078 Chr.15: 40853182-40853182 on Build GRCh38
rs201206539 Chr.15: 40853127-40853127 on Build GRCh38
rs769612807 Chr.15: 40853030-40853030 on Build GRCh38
rs545207208 Chr.15: 40853033-40853033 on Build GRCh38
rs774073564 Chr.15: 40853134-40853134 on Build GRCh38
rs372353456 Chr.15: 40853165-40853165 on Build GRCh38
rs763467419 Chr.15: 40853167-40853167 on Build GRCh38
rs371597596 Chr.15: 40853225-40853225 on Build GRCh38
rs1018812661 Chr.15: 40853205-40853206 on Build GRCh38
rs753395243 Chr.15: 40853198-40853198 on Build GRCh38
rs769976576 Chr.15: 40853150-40853150 on Build GRCh38
rs375870778 Chr.15: 40853168-40853168 on Build GRCh38
rs368407939 Chr.15: 40853164-40853164 on Build GRCh38
rs746728134 Chr.15: 40853226-40853226 on Build GRCh38
rs955844090 Chr.15: 40853200-40853200 on Build GRCh38
rs780307229 Chr.15: 40853145-40853145 on Build GRCh38
rs748175184 Chr.15: 40853051-40853051 on Build GRCh38
rs575604277 Chr.15: 40853055-40853055 on Build GRCh38
rs1012211965 Chr.15: 40853142-40853142 on Build GRCh38
rs139263716 Chr.15: 40853141-40853141 on Build GRCh38
rs778081820 Chr.15: 40853135-40853135 on Build GRCh38
rs1031525708 Chr.15: 40853171-40853171 on Build GRCh38
rs560850491 Chr.15: 40853098-40853098 on Build GRCh38
rs757233557 Chr.15: 40853118-40853118 on Build GRCh38
rs1003781706 Chr.15: 40853119-40853119 on Build GRCh38
rs901586136 Chr.15: 40853224-40853224 on Build GRCh38
rs934601176 Chr.15: 40853026-40853026 on Build GRCh38
rs367901148 Chr.15: 40853212-40853212 on Build GRCh38
rs768716685 Chr.15: 40853148-40853148 on Build GRCh38
rs202101923 Chr.15: 40853039-40853039 on Build GRCh38
rs756719173 Chr.15: 40853208-40853208 on Build GRCh38
rs760398562 Chr.15: 40853186-40853186 on Build GRCh38
rs377397915 Chr.15: 40853151-40853151 on Build GRCh38
rs544556664 Chr.15: 40853093-40853093 on Build GRCh38
rs759250856 Chr.15: 40853184-40853184 on Build GRCh38

View Full Product Details