Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10902732-10903001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYCP2L
Entrez Gene ID
221711
Gene Name
synaptonemal complex protein 2 like
Gene Aliases
C6orf177, NO145, dJ62D2.1
UniGene
Hs.729128
Gene Chromosome Location
Chr.6: 10886831-10974309 on Build GRCh38
Transcripts
NM_001040274.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6997416 c.553-2delA p.? Chr.6: 10902873-10902873 on Build GRCh38
7392554 c.568A>T p.N190Y Chr.6: 10902890-10902890 on Build GRCh38
6189187 c.637C>G p.L213V Chr.6: 10902959-10902959 on Build GRCh38
3619332 c.590C>T p.P197L Chr.6: 10902912-10902912 on Build GRCh38
3155064 c.610T>A p.F204I Chr.6: 10902932-10902932 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768545885 Chr.6: 10902853-10902853 on Build GRCh38
rs781782154 Chr.6: 10902904-10902904 on Build GRCh38
rs570590282 Chr.6: 10902819-10902819 on Build GRCh38
rs560332494 Chr.6: 10902831-10902831 on Build GRCh38
rs200655499 Chr.6: 10902920-10902920 on Build GRCh38
rs572341599 Chr.6: 10902843-10902843 on Build GRCh38
rs745891181 Chr.6: 10902966-10902966 on Build GRCh38
rs1046334492 Chr.6: 10902789-10902789 on Build GRCh38
rs753260209 Chr.6: 10902907-10902907 on Build GRCh38
rs768297339 Chr.6: 10902936-10902937 on Build GRCh38
rs777414730 Chr.6: 10902943-10902943 on Build GRCh38
rs771876505 Chr.6: 10902781-10902781 on Build GRCh38
rs747830755 Chr.6: 10902925-10902925 on Build GRCh38
rs757695405 Chr.6: 10902892-10902892 on Build GRCh38
rs770957451 Chr.6: 10902801-10902801 on Build GRCh38
rs770742324 Chr.6: 10902955-10902955 on Build GRCh38
rs764367600 Chr.6: 10902848-10902848 on Build GRCh38
rs374633930 Chr.6: 10902766-10902766 on Build GRCh38
rs543040307 Chr.6: 10902779-10902779 on Build GRCh38
rs762521102 Chr.6: 10902807-10902807 on Build GRCh38
rs999162607 Chr.6: 10902840-10902840 on Build GRCh38
rs760536511 Chr.6: 10902800-10902800 on Build GRCh38
rs768296347 Chr.6: 10902767-10902767 on Build GRCh38
rs542793014 Chr.6: 10902918-10902918 on Build GRCh38
rs369230080 Chr.6: 10902905-10902905 on Build GRCh38
rs752985725 Chr.6: 10902838-10902838 on Build GRCh38
rs752044331 Chr.6: 10902862-10902862 on Build GRCh38
rs757809869 Chr.6: 10902870-10902870 on Build GRCh38
rs776567340 Chr.6: 10902812-10902812 on Build GRCh38
rs758123953 Chr.6: 10902940-10902940 on Build GRCh38
rs778581066 Chr.6: 10902778-10902778 on Build GRCh38
rs375077118 Chr.6: 10902784-10902785 on Build GRCh38
rs776680477 Chr.6: 10902957-10902957 on Build GRCh38
rs746888180 Chr.6: 10902950-10902950 on Build GRCh38
rs765534097 Chr.6: 10902837-10902837 on Build GRCh38
rs545327866 Chr.6: 10902783-10902783 on Build GRCh38

View Full Product Details