Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 97307783-97308020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC180
Entrez Gene ID
100499483
Gene Name
coiled-coil domain containing 180
Gene Aliases
BDAG1, C9orf174, KIAA1529
UniGene
Hs.435629
Gene Chromosome Location
Chr.9: 97307628-97377295 on Build GRCh38
Transcripts
NM_020893.2 NR_036527.1 NR_036528.1 NR_036529.1

Gene Symbol
LOC100499484-C9ORF174
Entrez Gene ID
57653
Gene Name
LOC100499484-C9orf174 readthrough
Gene Aliases
-
UniGene
Hs.435629
Gene Chromosome Location
Chr.9: 97238426-97377295 on Build GRCh38
Transcripts
NM_020893.2 NR_036527.1 NR_036528.1 NR_036529.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6621939 c.501G>A p.A167A Chr.9: 97308015-97308015 on Build GRCh38
6621968 c.82G>A p.A28T Chr.9: 97308013-97308013 on Build GRCh38
4512056 c.88C>T p.P30S Chr.9: 97308019-97308019 on Build GRCh38
6621969 c.499G>A p.A167T Chr.9: 97308013-97308013 on Build GRCh38
3902383 c.79G>A p.D27N Chr.9: 97308010-97308010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537758184 Chr.9: 97307933-97307933 on Build GRCh38
rs370078752 Chr.9: 97307869-97307869 on Build GRCh38
rs769861146 Chr.9: 97307953-97307953 on Build GRCh38
rs938009299 Chr.9: 97307904-97307904 on Build GRCh38
rs767876516 Chr.9: 97307835-97307835 on Build GRCh38
rs749462552 Chr.9: 97307963-97307963 on Build GRCh38
rs751093296 Chr.9: 97307860-97307860 on Build GRCh38
rs535094962 Chr.9: 97307870-97307870 on Build GRCh38
rs939876437 Chr.9: 97307930-97307930 on Build GRCh38
rs761270682 Chr.9: 97307999-97307999 on Build GRCh38
rs375970784 Chr.9: 97307937-97307937 on Build GRCh38
rs986421117 Chr.9: 97307832-97307832 on Build GRCh38
rs771089629 Chr.9: 97307966-97307966 on Build GRCh38
rs184844600 Chr.9: 97307841-97307841 on Build GRCh38
rs909774855 Chr.9: 97307928-97307928 on Build GRCh38
rs1052575230 Chr.9: 97307826-97307826 on Build GRCh38
rs145635692 Chr.9: 97307978-97307978 on Build GRCh38
rs747008755 Chr.9: 97307946-97307946 on Build GRCh38
rs1048301616 Chr.9: 97307908-97307908 on Build GRCh38
rs374283901 Chr.9: 97307861-97307861 on Build GRCh38
rs759794148 Chr.9: 97307985-97307985 on Build GRCh38
rs760245890 Chr.9: 97307850-97307850 on Build GRCh38
rs377488090 Chr.9: 97307855-97307855 on Build GRCh38
rs756789028 Chr.9: 97307889-97307889 on Build GRCh38
rs546247608 Chr.9: 97307877-97307877 on Build GRCh38
rs763642707 Chr.9: 97308000-97308000 on Build GRCh38
rs112001128 Chr.9: 97307824-97307824 on Build GRCh38
rs778724968 Chr.9: 97307935-97307935 on Build GRCh38
rs779834161 Chr.9: 97307941-97307941 on Build GRCh38
rs776133596 Chr.9: 97307990-97307990 on Build GRCh38
rs371777549 Chr.9: 97307857-97307857 on Build GRCh38
rs1025399047 Chr.9: 97307886-97307886 on Build GRCh38
rs373893600 Chr.9: 97307825-97307825 on Build GRCh38
rs368770897 Chr.9: 97307962-97307962 on Build GRCh38
rs971241081 Chr.9: 97307866-97307866 on Build GRCh38
rs559160380 Chr.9: 97307948-97307948 on Build GRCh38
rs765835406 Chr.9: 97307829-97307829 on Build GRCh38
rs556963062 Chr.9: 97307871-97307871 on Build GRCh38
rs772542890 Chr.9: 97307986-97307986 on Build GRCh38

View Full Product Details