Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 19142941-19143166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC13
Entrez Gene ID
54503
Gene Name
zinc finger DHHC-type containing 13
Gene Aliases
HIP14L, HIP3RP
UniGene
Hs.188569
Gene Chromosome Location
Chr.11: 19117099-19176420 on Build GRCh38
Transcripts
NM_001001483.2 NM_019028.2 XM_005252996.2 XM_011520194.2 XM_011520195.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2093216 c.51C>T p.P17P Chr.11: 19143001-19143001 on Build GRCh38
2093220 c.167C>T p.A56V Chr.11: 19143117-19143117 on Build GRCh38
1585668 c.118G>T p.E40* Chr.11: 19143068-19143068 on Build GRCh38
1585667 c.151_152delTG p.C51fs*1 Chr.11: 19143101-19143102 on Build GRCh38
4031985 c.104T>C p.L35P Chr.11: 19143054-19143054 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748136141 Chr.11: 19143045-19143045 on Build GRCh38
rs369100465 Chr.11: 19143012-19143012 on Build GRCh38
rs555075710 Chr.11: 19142991-19142991 on Build GRCh38
rs781222431 Chr.11: 19143095-19143095 on Build GRCh38
rs759437411 Chr.11: 19143053-19143053 on Build GRCh38
rs927476965 Chr.11: 19143004-19143004 on Build GRCh38
rs891705502 Chr.11: 19143097-19143097 on Build GRCh38
rs780563238 Chr.11: 19143025-19143025 on Build GRCh38
rs200413197 Chr.11: 19143123-19143123 on Build GRCh38
rs750023488 Chr.11: 19143069-19143069 on Build GRCh38
rs747437838 Chr.11: 19143034-19143034 on Build GRCh38
rs375837665 Chr.11: 19142967-19142967 on Build GRCh38
rs774302722 Chr.11: 19142980-19142980 on Build GRCh38
rs1031577150 Chr.11: 19143079-19143079 on Build GRCh38
rs373647126 Chr.11: 19143020-19143020 on Build GRCh38
rs781481593 Chr.11: 19143041-19143041 on Build GRCh38
rs748378291 Chr.11: 19143106-19143106 on Build GRCh38
rs542324220 Chr.11: 19143010-19143010 on Build GRCh38
rs748690221 Chr.11: 19143123-19143126 on Build GRCh38
rs769948653 Chr.11: 19143110-19143110 on Build GRCh38
rs199613321 Chr.11: 19143058-19143058 on Build GRCh38
rs759276019 Chr.11: 19142982-19142982 on Build GRCh38
rs760527717 Chr.11: 19143061-19143061 on Build GRCh38
rs777823589 Chr.11: 19143119-19143119 on Build GRCh38
rs753554311 Chr.11: 19143065-19143065 on Build GRCh38
rs780722559 Chr.11: 19143084-19143084 on Build GRCh38
rs759017825 Chr.11: 19143024-19143024 on Build GRCh38
rs763583737 Chr.11: 19143062-19143062 on Build GRCh38
rs768702628 Chr.11: 19143038-19143038 on Build GRCh38
rs761425439 Chr.11: 19143067-19143067 on Build GRCh38
rs755756854 Chr.11: 19142998-19142998 on Build GRCh38
rs1016166455 Chr.11: 19143074-19143074 on Build GRCh38
rs758961008 Chr.11: 19143080-19143080 on Build GRCh38
rs202238805 Chr.11: 19143046-19143046 on Build GRCh38
rs773327835 Chr.11: 19143050-19143050 on Build GRCh38
rs752525406 Chr.11: 19142994-19142994 on Build GRCh38
rs1056473256 Chr.11: 19143003-19143003 on Build GRCh38
rs139128805 Chr.11: 19143022-19143022 on Build GRCh38
rs376906312 Chr.11: 19143021-19143021 on Build GRCh38
rs776560974 Chr.11: 19142972-19142972 on Build GRCh38
rs768784588 Chr.11: 19142976-19142976 on Build GRCh38
rs755400918 Chr.11: 19143092-19143092 on Build GRCh38
rs761966669 Chr.11: 19142975-19142975 on Build GRCh38
rs763861295 Chr.11: 19142999-19142999 on Build GRCh38
rs572710864 Chr.11: 19143068-19143068 on Build GRCh38

View Full Product Details