Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 71698699-71698930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM19
Entrez Gene ID
55266
Gene Name
transmembrane protein 19
Gene Aliases
-
UniGene
Hs.644802
Gene Chromosome Location
Chr.12: 71686098-71704060 on Build GRCh38
Transcripts
NM_018279.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
695165 c.649G>A p.G217R Chr.12: 71698911-71698911 on Build GRCh38
5462270 c.657A>G p.T219T Chr.12: 71698919-71698919 on Build GRCh38
5462271 c.657A>G p.T219T Chr.12: 71698919-71698919 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs914578238 Chr.12: 71698802-71698802 on Build GRCh38
rs775260803 Chr.12: 71698873-71698873 on Build GRCh38
rs750043573 Chr.12: 71698866-71698866 on Build GRCh38
rs919921272 Chr.12: 71698823-71698823 on Build GRCh38
rs563539606 Chr.12: 71698839-71698839 on Build GRCh38
rs775916622 Chr.12: 71698877-71698877 on Build GRCh38
rs773257046 Chr.12: 71698871-71698871 on Build GRCh38
rs961927544 Chr.12: 71698732-71698732 on Build GRCh38
rs182858452 Chr.12: 71698846-71698846 on Build GRCh38
rs528050405 Chr.12: 71698739-71698739 on Build GRCh38
rs749524172 Chr.12: 71698872-71698872 on Build GRCh38
rs533715795 Chr.12: 71698734-71698734 on Build GRCh38
rs373974534 Chr.12: 71698807-71698811 on Build GRCh38
rs763913167 Chr.12: 71698907-71698907 on Build GRCh38
rs947373178 Chr.12: 71698831-71698831 on Build GRCh38
rs760807483 Chr.12: 71698883-71698883 on Build GRCh38
rs573165166 Chr.12: 71698804-71698808 on Build GRCh38
rs549228941 Chr.12: 71698906-71698906 on Build GRCh38
rs776552763 Chr.12: 71698894-71698895 on Build GRCh38
rs776667945 Chr.12: 71698908-71698908 on Build GRCh38
rs972761759 Chr.12: 71698740-71698740 on Build GRCh38
rs1055852786 Chr.12: 71698762-71698762 on Build GRCh38
rs1041698268 Chr.12: 71698883-71698883 on Build GRCh38
rs985527601 Chr.12: 71698844-71698844 on Build GRCh38
rs369382884 Chr.12: 71698870-71698870 on Build GRCh38
rs113317309 Chr.12: 71698738-71698738 on Build GRCh38
rs931408211 Chr.12: 71698835-71698835 on Build GRCh38
rs772439140 Chr.12: 71698871-71698871 on Build GRCh38
rs768820736 Chr.12: 71698885-71698886 on Build GRCh38
rs902731935 Chr.12: 71698885-71698885 on Build GRCh38
rs375156115 Chr.12: 71698856-71698856 on Build GRCh38
rs7980112 Chr.12: 71698731-71698731 on Build GRCh38

View Full Product Details