Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 39572302-39572545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM14
Entrez Gene ID
201895
Gene Name
small integral membrane protein 14
Gene Aliases
C4orf34
UniGene
Hs.205952
Gene Chromosome Location
Chr.4: 39547297-39638998 on Build GRCh38
Transcripts
NM_001317896.1 NM_001317897.1 NM_174921.2 XM_011513640.2 XM_017007862.1 XM_017007863.1 NR_047679.1

Gene Symbol
UGDH-AS1
Entrez Gene ID
100885776
Gene Name
UGDH antisense RNA 1
Gene Aliases
-
UniGene
Hs.640769
Gene Chromosome Location
Chr.4: 39527839-39594707 on Build GRCh38
Transcripts
NM_001317896.1 NM_001317897.1 NM_174921.2 XM_011513640.2 XM_017007862.1 XM_017007863.1 NR_047679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3946341 c.79C>T p.R27W Chr.4: 39572460-39572460 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs535826625 Chr.4: 39572511-39572511 on Build GRCh38
rs192328356 Chr.4: 39572349-39572349 on Build GRCh38
rs763880366 Chr.4: 39572379-39572379 on Build GRCh38
rs961900468 Chr.4: 39572398-39572398 on Build GRCh38
rs757011123 Chr.4: 39572446-39572446 on Build GRCh38
rs777620115 Chr.4: 39572399-39572400 on Build GRCh38
rs567268237 Chr.4: 39572490-39572490 on Build GRCh38
rs778521151 Chr.4: 39572494-39572499 on Build GRCh38
rs748858271 Chr.4: 39572501-39572501 on Build GRCh38
rs1028621993 Chr.4: 39572385-39572385 on Build GRCh38
rs745814949 Chr.4: 39572484-39572484 on Build GRCh38
rs779166318 Chr.4: 39572428-39572428 on Build GRCh38
rs780134707 Chr.4: 39572415-39572415 on Build GRCh38
rs769446976 Chr.4: 39572372-39572372 on Build GRCh38
rs762149069 Chr.4: 39572386-39572386 on Build GRCh38
rs753956390 Chr.4: 39572419-39572419 on Build GRCh38
rs748400883 Chr.4: 39572441-39572441 on Build GRCh38
rs189335228 Chr.4: 39572375-39572375 on Build GRCh38
rs369341511 Chr.4: 39572381-39572381 on Build GRCh38
rs756594520 Chr.4: 39572409-39572409 on Build GRCh38
rs756749839 Chr.4: 39572489-39572508 on Build GRCh38
rs780987245 Chr.4: 39572469-39572469 on Build GRCh38
rs750907676 Chr.4: 39572400-39572400 on Build GRCh38
rs892129356 Chr.4: 39572473-39572473 on Build GRCh38
rs753779669 Chr.4: 39572437-39572438 on Build GRCh38
rs775538619 Chr.4: 39572493-39572493 on Build GRCh38
rs372764933 Chr.4: 39572377-39572377 on Build GRCh38
rs767855530 Chr.4: 39572393-39572393 on Build GRCh38
rs184755694 Chr.4: 39572373-39572373 on Build GRCh38
rs564672500 Chr.4: 39572352-39572352 on Build GRCh38
rs755025674 Chr.4: 39572426-39572426 on Build GRCh38

View Full Product Details