Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 101177686-101177929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHSY1
Entrez Gene ID
22856
Gene Name
chondroitin sulfate synthase 1
Gene Aliases
CHSY, CSS1, ChSy-1, TPBS
UniGene
Hs.110488
Gene Chromosome Location
Chr.15: 101175723-101251932 on Build GRCh38
Transcripts
NM_014918.4 XM_006720435.3 XM_011521364.2 XM_017022011.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4419068 c.1896C>T p.V632V Chr.15: 101177901-101177901 on Build GRCh38
4799190 c.2092A>G p.I698V Chr.15: 101177705-101177705 on Build GRCh38
6141641 c.1915G>C p.E639Q Chr.15: 101177882-101177882 on Build GRCh38
3815691 c.1897G>C p.D633H Chr.15: 101177900-101177900 on Build GRCh38
5628038 c.2062T>A p.W688R Chr.15: 101177735-101177735 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896474550 Chr.15: 101177784-101177784 on Build GRCh38
rs756636905 Chr.15: 101177754-101177754 on Build GRCh38
rs778446867 Chr.15: 101177767-101177767 on Build GRCh38
rs762093887 Chr.15: 101177730-101177730 on Build GRCh38
rs979729208 Chr.15: 101177839-101177839 on Build GRCh38
rs4426333 Chr.15: 101177841-101177841 on Build GRCh38
rs753396162 Chr.15: 101177758-101177758 on Build GRCh38
rs3803423 Chr.15: 101177901-101177901 on Build GRCh38
rs145837770 Chr.15: 101177757-101177757 on Build GRCh38
rs570222707 Chr.15: 101177825-101177825 on Build GRCh38
rs767851737 Chr.15: 101177737-101177737 on Build GRCh38
rs368276647 Chr.15: 101177904-101177904 on Build GRCh38
rs758159379 Chr.15: 101177776-101177776 on Build GRCh38
rs968677826 Chr.15: 101177778-101177778 on Build GRCh38
rs764764707 Chr.15: 101177895-101177895 on Build GRCh38
rs754721596 Chr.15: 101177761-101177761 on Build GRCh38
rs771860435 Chr.15: 101177785-101177785 on Build GRCh38
rs774436935 Chr.15: 101177835-101177835 on Build GRCh38
rs773135273 Chr.15: 101177791-101177791 on Build GRCh38
rs770823445 Chr.15: 101177822-101177822 on Build GRCh38
rs758974494 Chr.15: 101177894-101177894 on Build GRCh38
rs949389424 Chr.15: 101177789-101177789 on Build GRCh38
rs750997098 Chr.15: 101177745-101177745 on Build GRCh38
rs777577489 Chr.15: 101177783-101177783 on Build GRCh38
rs896654554 Chr.15: 101177743-101177743 on Build GRCh38
rs150075110 Chr.15: 101177840-101177840 on Build GRCh38
rs747770781 Chr.15: 101177775-101177775 on Build GRCh38
rs558201715 Chr.15: 101177788-101177788 on Build GRCh38
rs961461120 Chr.15: 101177899-101177899 on Build GRCh38
rs773686281 Chr.15: 101177843-101177843 on Build GRCh38
rs752317596 Chr.15: 101177900-101177900 on Build GRCh38
rs1009725006 Chr.15: 101177860-101177860 on Build GRCh38
rs371386152 Chr.15: 101177903-101177903 on Build GRCh38
rs747715647 Chr.15: 101177768-101177768 on Build GRCh38
rs141799040 Chr.15: 101177837-101177837 on Build GRCh38
rs372896966 Chr.15: 101177826-101177826 on Build GRCh38
rs28364839 Chr.15: 101177892-101177892 on Build GRCh38
rs375906150 Chr.15: 101177863-101177863 on Build GRCh38
rs768722457 Chr.15: 101177832-101177832 on Build GRCh38
rs1034764038 Chr.15: 101177725-101177725 on Build GRCh38
rs776452925 Chr.15: 101177842-101177842 on Build GRCh38
rs761334027 Chr.15: 101177858-101177858 on Build GRCh38

View Full Product Details