Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 64279951-64280208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TECRL
Entrez Gene ID
253017
Gene Name
trans-2,3-enoyl-CoA reductase-like
Gene Aliases
GPSN2L, SRD5A2L2, TERL
UniGene
Hs.227752
Gene Chromosome Location
Chr.4: 64276298-64409509 on Build GRCh38
Transcripts
NM_001010874.4 XM_005265662.4 XM_005265663.2 XM_005265664.2 XM_005265665.3 XM_017007960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7189267 c.1030T>C p.Y344H Chr.4: 64280134-64280134 on Build GRCh38
3917998 c.1017G>A p.K339K Chr.4: 64280147-64280147 on Build GRCh38
1430519 c.1073T>C p.M358T Chr.4: 64280091-64280091 on Build GRCh38
4409010 c.1061G>T p.R354I Chr.4: 64280103-64280103 on Build GRCh38
7189251 c.1033C>A p.L345M Chr.4: 64280131-64280131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780720827 Chr.4: 64280166-64280166 on Build GRCh38
rs781650913 Chr.4: 64280159-64280159 on Build GRCh38
rs764218341 Chr.4: 64280155-64280155 on Build GRCh38
rs757538239 Chr.4: 64280158-64280158 on Build GRCh38
rs941074312 Chr.4: 64280069-64280069 on Build GRCh38
rs967413425 Chr.4: 64280006-64280006 on Build GRCh38
rs542128095 Chr.4: 64280041-64280041 on Build GRCh38
rs544073941 Chr.4: 64280018-64280018 on Build GRCh38
rs758477637 Chr.4: 64280151-64280151 on Build GRCh38
rs1011677840 Chr.4: 64279995-64279995 on Build GRCh38
rs946083598 Chr.4: 64280101-64280101 on Build GRCh38
rs555279932 Chr.4: 64280004-64280004 on Build GRCh38
rs761513125 Chr.4: 64280095-64280095 on Build GRCh38
rs777981538 Chr.4: 64280023-64280023 on Build GRCh38
rs145001397 Chr.4: 64280129-64280129 on Build GRCh38
rs757706380 Chr.4: 64280034-64280034 on Build GRCh38
rs775102562 Chr.4: 64280067-64280067 on Build GRCh38
rs1016689002 Chr.4: 64280025-64280025 on Build GRCh38
rs752713102 Chr.4: 64280137-64280137 on Build GRCh38
rs747269622 Chr.4: 64280026-64280026 on Build GRCh38
rs369830622 Chr.4: 64280065-64280065 on Build GRCh38
rs745969657 Chr.4: 64280067-64280067 on Build GRCh38
rs867567601 Chr.4: 64280127-64280127 on Build GRCh38
rs547241447 Chr.4: 64280040-64280040 on Build GRCh38
rs373062805 Chr.4: 64280091-64280091 on Build GRCh38
rs772239502 Chr.4: 64280140-64280141 on Build GRCh38
rs748744052 Chr.4: 64280173-64280173 on Build GRCh38
rs549574336 Chr.4: 64280161-64280161 on Build GRCh38
rs529545787 Chr.4: 64280132-64280132 on Build GRCh38
rs768366197 Chr.4: 64280088-64280088 on Build GRCh38
rs975352659 Chr.4: 64280051-64280051 on Build GRCh38
rs541617172 Chr.4: 64279991-64279991 on Build GRCh38
rs767405714 Chr.4: 64280104-64280104 on Build GRCh38
rs781535610 Chr.4: 64280036-64280036 on Build GRCh38
rs773003644 Chr.4: 64280123-64280123 on Build GRCh38
rs1046387190 Chr.4: 64280126-64280126 on Build GRCh38
rs564341054 Chr.4: 64280021-64280021 on Build GRCh38
rs751864841 Chr.4: 64280157-64280157 on Build GRCh38
rs748829708 Chr.4: 64280075-64280075 on Build GRCh38
rs954718269 Chr.4: 64280066-64280066 on Build GRCh38
rs575173950 Chr.4: 64280009-64280009 on Build GRCh38
rs768897184 Chr.4: 64280041-64280041 on Build GRCh38

View Full Product Details