Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54991693-54991954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM61
Entrez Gene ID
199964
Gene Name
transmembrane protein 61
Gene Aliases
-
UniGene
Hs.663950
Gene Chromosome Location
Chr.1: 54980614-54992293 on Build GRCh38
Transcripts
NM_182532.2 XM_005270586.4 XM_011540911.2 XM_011540912.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1737063 c.454A>T p.T152S Chr.1: 54991924-54991924 on Build GRCh38
5381277 c.423C>T p.A141A Chr.1: 54991893-54991893 on Build GRCh38
7103651 c.380T>C p.V127A Chr.1: 54991850-54991850 on Build GRCh38
5540541 c.459G>A p.E153E Chr.1: 54991929-54991929 on Build GRCh38
4009008 c.397G>A p.E133K Chr.1: 54991867-54991867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs545409931 Chr.1: 54991731-54991731 on Build GRCh38
rs771668298 Chr.1: 54991910-54991910 on Build GRCh38
rs747101338 Chr.1: 54991807-54991807 on Build GRCh38
rs751628640 Chr.1: 54991878-54991878 on Build GRCh38
rs777163589 Chr.1: 54991820-54991820 on Build GRCh38
rs200626777 Chr.1: 54991876-54991876 on Build GRCh38
rs766261829 Chr.1: 54991859-54991859 on Build GRCh38
rs373373486 Chr.1: 54991792-54991792 on Build GRCh38
rs998486055 Chr.1: 54991881-54991881 on Build GRCh38
rs979147431 Chr.1: 54991850-54991850 on Build GRCh38
rs757206853 Chr.1: 54991884-54991884 on Build GRCh38
rs369985259 Chr.1: 54991786-54991786 on Build GRCh38
rs780458709 Chr.1: 54991899-54991899 on Build GRCh38
rs139141549 Chr.1: 54991916-54991916 on Build GRCh38
rs562074103 Chr.1: 54991860-54991860 on Build GRCh38
rs556351250 Chr.1: 54991757-54991757 on Build GRCh38
rs201179249 Chr.1: 54991913-54991913 on Build GRCh38
rs761385381 Chr.1: 54991835-54991835 on Build GRCh38
rs781572304 Chr.1: 54991891-54991891 on Build GRCh38
rs771746688 Chr.1: 54991840-54991840 on Build GRCh38
rs201375691 Chr.1: 54991894-54991894 on Build GRCh38
rs201950532 Chr.1: 54991793-54991793 on Build GRCh38
rs367741541 Chr.1: 54991866-54991866 on Build GRCh38
rs746261344 Chr.1: 54991821-54991821 on Build GRCh38
rs374480075 Chr.1: 54991826-54991826 on Build GRCh38
rs772652696 Chr.1: 54991844-54991844 on Build GRCh38
rs754782335 Chr.1: 54991790-54991790 on Build GRCh38
rs770095837 Chr.1: 54991823-54991823 on Build GRCh38
rs144935772 Chr.1: 54991921-54991921 on Build GRCh38
rs746314762 Chr.1: 54991893-54991893 on Build GRCh38
rs142193282 Chr.1: 54991907-54991907 on Build GRCh38
rs377259600 Chr.1: 54991900-54991900 on Build GRCh38
rs765253556 Chr.1: 54991936-54991936 on Build GRCh38
rs758670966 Chr.1: 54991838-54991838 on Build GRCh38
rs539683043 Chr.1: 54991738-54991738 on Build GRCh38
rs750469834 Chr.1: 54991797-54991799 on Build GRCh38
rs760206795 Chr.1: 54991854-54991854 on Build GRCh38
rs142872695 Chr.1: 54991867-54991867 on Build GRCh38
rs771392374 Chr.1: 54991816-54991816 on Build GRCh38
rs759615637 Chr.1: 54991935-54991935 on Build GRCh38
rs758753301 Chr.1: 54991875-54991875 on Build GRCh38
rs181018102 Chr.1: 54991925-54991925 on Build GRCh38

View Full Product Details