Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 64213021-64213268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF735
Entrez Gene ID
730291
Gene Name
zinc finger protein 735
Gene Aliases
ZNF735P
UniGene
Hs.723116
Gene Chromosome Location
Chr.7: 64207203-64220290 on Build GRCh38
Transcripts
NM_001159524.1

SNP Detail

SNP ID Chromosome Location
rs758057746 Chr.7: 64213112-64213112 on Build GRCh38
rs113133873 Chr.7: 64213225-64213225 on Build GRCh38
rs767684801 Chr.7: 64213159-64213159 on Build GRCh38
rs746463609 Chr.7: 64213056-64213056 on Build GRCh38
rs749684455 Chr.7: 64213107-64213107 on Build GRCh38
rs746692812 Chr.7: 64213131-64213131 on Build GRCh38
rs764568122 Chr.7: 64213173-64213173 on Build GRCh38
rs754662775 Chr.7: 64213089-64213089 on Build GRCh38
rs779272305 Chr.7: 64213179-64213179 on Build GRCh38
rs761242701 Chr.7: 64213170-64213170 on Build GRCh38
rs769682219 Chr.7: 64213136-64213136 on Build GRCh38
rs749282296 Chr.7: 64213217-64213217 on Build GRCh38
rs10252019 Chr.7: 64213230-64213230 on Build GRCh38
rs915489620 Chr.7: 64213219-64213219 on Build GRCh38
rs985608339 Chr.7: 64213215-64213215 on Build GRCh38
rs751189655 Chr.7: 64213196-64213196 on Build GRCh38
rs763229737 Chr.7: 64213079-64213079 on Build GRCh38
rs760245280 Chr.7: 64213078-64213078 on Build GRCh38
rs752865023 Chr.7: 64213169-64213169 on Build GRCh38
rs751847673 Chr.7: 64213084-64213084 on Build GRCh38
rs779613441 Chr.7: 64213119-64213119 on Build GRCh38
rs776511099 Chr.7: 64213063-64213063 on Build GRCh38
rs765756884 Chr.7: 64213077-64213078 on Build GRCh38
rs117854459 Chr.7: 64213103-64213103 on Build GRCh38
rs768157282 Chr.7: 64213058-64213058 on Build GRCh38
rs780672907 Chr.7: 64213201-64213201 on Build GRCh38
rs773044727 Chr.7: 64213139-64213139 on Build GRCh38
rs767740434 Chr.7: 64213093-64213093 on Build GRCh38
rs753518901 Chr.7: 64213078-64213079 on Build GRCh38
rs761761363 Chr.7: 64213066-64213066 on Build GRCh38
rs954397858 Chr.7: 64213213-64213213 on Build GRCh38
rs769199705 Chr.7: 64213126-64213127 on Build GRCh38
rs774677555 Chr.7: 64213155-64213155 on Build GRCh38
rs894057195 Chr.7: 64213097-64213097 on Build GRCh38
rs1012744123 Chr.7: 64213164-64213164 on Build GRCh38
rs551760886 Chr.7: 64213083-64213083 on Build GRCh38
rs753210390 Chr.7: 64213094-64213094 on Build GRCh38
rs1050459099 Chr.7: 64213088-64213088 on Build GRCh38
rs766716718 Chr.7: 64213199-64213199 on Build GRCh38
rs765055114 Chr.7: 64213070-64213070 on Build GRCh38
rs759863714 Chr.7: 64213157-64213157 on Build GRCh38
rs757520449 Chr.7: 64213178-64213178 on Build GRCh38
rs772489501 Chr.7: 64213234-64213234 on Build GRCh38
rs762594074 Chr.7: 64213151-64213151 on Build GRCh38
rs770767417 Chr.7: 64213152-64213152 on Build GRCh38
rs772917483 Chr.7: 64213071-64213071 on Build GRCh38
rs545330625 Chr.7: 64213208-64213208 on Build GRCh38
rs183236398 Chr.7: 64213135-64213135 on Build GRCh38
rs529434514 Chr.7: 64213209-64213209 on Build GRCh38
rs755044812 Chr.7: 64213091-64213091 on Build GRCh38
rs754372617 Chr.7: 64213174-64213174 on Build GRCh38
rs747667984 Chr.7: 64213205-64213205 on Build GRCh38

View Full Product Details