Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44678770-44678999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM19
Entrez Gene ID
56971
Gene Name
carcinoembryonic antigen related cell adhesion molecule 19
Gene Aliases
CEACM19, CEAL1
UniGene
Hs.416925
Gene Chromosome Location
Chr.19: 44671452-44684355 on Build GRCh38
Transcripts
NM_001127893.1 NM_020219.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6198904 c.576-7A>C p.? Chr.19: 44678846-44678846 on Build GRCh38
3727606 c.659+2T>C p.? Chr.19: 44678938-44678938 on Build GRCh38
3535702 c.602C>T p.S201F Chr.19: 44678879-44678879 on Build GRCh38
3707209 c.604C>G p.L202V Chr.19: 44678881-44678881 on Build GRCh38
1394487 c.648G>A p.T216T Chr.19: 44678925-44678925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930049953 Chr.19: 44678893-44678893 on Build GRCh38
rs767123178 Chr.19: 44678893-44678893 on Build GRCh38
rs766087535 Chr.19: 44678914-44678914 on Build GRCh38
rs768597256 Chr.19: 44678871-44678871 on Build GRCh38
rs753915476 Chr.19: 44678942-44678942 on Build GRCh38
rs762712982 Chr.19: 44678910-44678910 on Build GRCh38
rs1001209032 Chr.19: 44678968-44678968 on Build GRCh38
rs193089064 Chr.19: 44678822-44678822 on Build GRCh38
rs140104524 Chr.19: 44678898-44678898 on Build GRCh38
rs546697977 Chr.19: 44678865-44678865 on Build GRCh38
rs757422349 Chr.19: 44678838-44678838 on Build GRCh38
rs763418819 Chr.19: 44678804-44678804 on Build GRCh38
rs751920470 Chr.19: 44678818-44678818 on Build GRCh38
rs772963138 Chr.19: 44678902-44678902 on Build GRCh38
rs367678406 Chr.19: 44678934-44678934 on Build GRCh38
rs867137224 Chr.19: 44678962-44678962 on Build GRCh38
rs749439278 Chr.19: 44678835-44678835 on Build GRCh38
rs768194567 Chr.19: 44678925-44678925 on Build GRCh38
rs756132840 Chr.19: 44678833-44678833 on Build GRCh38
rs776630929 Chr.19: 44678924-44678924 on Build GRCh38
rs779857172 Chr.19: 44678864-44678864 on Build GRCh38
rs946597456 Chr.19: 44678855-44678855 on Build GRCh38
rs778124151 Chr.19: 44678834-44678834 on Build GRCh38
rs373826358 Chr.19: 44678878-44678878 on Build GRCh38
rs760847539 Chr.19: 44678937-44678937 on Build GRCh38
rs757477258 Chr.19: 44678964-44678964 on Build GRCh38
rs761339726 Chr.19: 44678837-44678838 on Build GRCh38
rs935148699 Chr.19: 44678820-44678820 on Build GRCh38
rs1006936415 Chr.19: 44678913-44678913 on Build GRCh38
rs774857502 Chr.19: 44678915-44678915 on Build GRCh38
rs566884681 Chr.19: 44678889-44678889 on Build GRCh38
rs200616815 Chr.19: 44678823-44678823 on Build GRCh38
rs770836274 Chr.19: 44678814-44678814 on Build GRCh38
rs751059898 Chr.19: 44678802-44678803 on Build GRCh38
rs903734764 Chr.19: 44678935-44678935 on Build GRCh38

View Full Product Details