Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 17117783-17118018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XYLT1
Entrez Gene ID
64131
Gene Name
xylosyltransferase 1
Gene Aliases
DBQD2, PXYLT1, XT-I, XT1, XTI, XYLTI, xylT-I
UniGene
Hs.22907
Gene Chromosome Location
Chr.16: 17102324-17470934 on Build GRCh38
Transcripts
NM_022166.3 XM_017023539.1 XM_017023540.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1197743 c.2353G>T p.V785L Chr.16: 17117850-17117850 on Build GRCh38
4388442 c.2273G>A p.G758D Chr.16: 17117930-17117930 on Build GRCh38
2127772 c.2261G>T p.R754L Chr.16: 17117942-17117942 on Build GRCh38
3506497 c.2285C>T p.P762L Chr.16: 17117918-17117918 on Build GRCh38
3420792 c.2410G>A p.E804K Chr.16: 17117793-17117793 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748617234 Chr.16: 17117969-17117969 on Build GRCh38
rs376063818 Chr.16: 17117906-17117906 on Build GRCh38
rs757011703 Chr.16: 17117850-17117850 on Build GRCh38
rs778581218 Chr.16: 17117859-17117859 on Build GRCh38
rs771245911 Chr.16: 17117985-17117985 on Build GRCh38
rs762255466 Chr.16: 17117804-17117804 on Build GRCh38
rs779548774 Chr.16: 17117866-17117866 on Build GRCh38
rs12325442 Chr.16: 17117991-17117991 on Build GRCh38
rs374020411 Chr.16: 17117943-17117943 on Build GRCh38
rs761614484 Chr.16: 17117926-17117926 on Build GRCh38
rs748564029 Chr.16: 17117875-17117875 on Build GRCh38
rs201348549 Chr.16: 17117841-17117841 on Build GRCh38
rs769344701 Chr.16: 17117869-17117869 on Build GRCh38
rs947041573 Chr.16: 17117837-17117837 on Build GRCh38
rs12325439 Chr.16: 17117907-17117907 on Build GRCh38
rs911941636 Chr.16: 17117848-17117848 on Build GRCh38
rs1021184322 Chr.16: 17117987-17117987 on Build GRCh38
rs762406815 Chr.16: 17117857-17117858 on Build GRCh38
rs145121977 Chr.16: 17117916-17117916 on Build GRCh38
rs770314311 Chr.16: 17117976-17117976 on Build GRCh38
rs753694444 Chr.16: 17117847-17117847 on Build GRCh38
rs753749607 Chr.16: 17117923-17117923 on Build GRCh38
rs758010563 Chr.16: 17117936-17117936 on Build GRCh38
rs74750647 Chr.16: 17117833-17117833 on Build GRCh38
rs139693964 Chr.16: 17117977-17117977 on Build GRCh38
rs765782913 Chr.16: 17117987-17117988 on Build GRCh38
rs906275352 Chr.16: 17117931-17117931 on Build GRCh38
rs766434865 Chr.16: 17117829-17117829 on Build GRCh38
rs142486601 Chr.16: 17117884-17117884 on Build GRCh38
rs764969137 Chr.16: 17117928-17117928 on Build GRCh38
rs774436763 Chr.16: 17117988-17117988 on Build GRCh38
rs1048279301 Chr.16: 17117893-17117893 on Build GRCh38
rs759525034 Chr.16: 17117910-17117910 on Build GRCh38
rs781777176 Chr.16: 17117951-17117951 on Build GRCh38
rs777076989 Chr.16: 17117803-17117803 on Build GRCh38
rs145615486 Chr.16: 17117845-17117845 on Build GRCh38
rs549518869 Chr.16: 17117871-17117871 on Build GRCh38
rs139558627 Chr.16: 17117818-17117818 on Build GRCh38
rs369216955 Chr.16: 17117825-17117825 on Build GRCh38
rs750045412 Chr.16: 17117930-17117930 on Build GRCh38
rs7201590 Chr.16: 17117872-17117872 on Build GRCh38
rs749488472 Chr.16: 17117983-17117983 on Build GRCh38
rs555809214 Chr.16: 17117942-17117942 on Build GRCh38
rs181524912 Chr.16: 17117865-17117865 on Build GRCh38
rs568680100 Chr.16: 17117881-17117881 on Build GRCh38
rs750040372 Chr.16: 17117860-17117860 on Build GRCh38
rs772086951 Chr.16: 17117993-17117993 on Build GRCh38
rs146457574 Chr.16: 17117905-17117905 on Build GRCh38
rs111893236 Chr.16: 17117827-17117827 on Build GRCh38
rs765621199 Chr.16: 17117811-17117811 on Build GRCh38

View Full Product Details