Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 155852118-155852389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC5
Entrez Gene ID
51802
Gene Name
acid sensing ion channel subunit family member 5
Gene Aliases
ACCN5, HINAC, INAC
UniGene
Hs.381349
Gene Chromosome Location
Chr.4: 155829699-155867303 on Build GRCh38
Transcripts
NM_017419.2 XM_017008291.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1052539 c.646C>A p.L216I Chr.4: 155852256-155852256 on Build GRCh38
4518904 c.586-1_586GG>AA p.? Chr.4: 155852316-155852317 on Build GRCh38
3601297 c.614G>A p.G205E Chr.4: 155852288-155852288 on Build GRCh38
3601298 c.586-1G>A p.? Chr.4: 155852317-155852317 on Build GRCh38
733355 c.653C>G p.A218G Chr.4: 155852249-155852249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919453348 Chr.4: 155852362-155852362 on Build GRCh38
rs757564895 Chr.4: 155852355-155852355 on Build GRCh38
rs771300037 Chr.4: 155852186-155852186 on Build GRCh38
rs772326049 Chr.4: 155852365-155852365 on Build GRCh38
rs781768790 Chr.4: 155852147-155852147 on Build GRCh38
rs771099825 Chr.4: 155852166-155852166 on Build GRCh38
rs200759770 Chr.4: 155852262-155852262 on Build GRCh38
rs145168162 Chr.4: 155852254-155852254 on Build GRCh38
rs754543137 Chr.4: 155852227-155852227 on Build GRCh38
rs200244304 Chr.4: 155852325-155852325 on Build GRCh38
rs149216368 Chr.4: 155852194-155852194 on Build GRCh38
rs991258784 Chr.4: 155852313-155852313 on Build GRCh38
rs958536874 Chr.4: 155852310-155852310 on Build GRCh38
rs137858552 Chr.4: 155852269-155852269 on Build GRCh38
rs746021780 Chr.4: 155852270-155852270 on Build GRCh38
rs201426530 Chr.4: 155852231-155852231 on Build GRCh38
rs769655464 Chr.4: 155852309-155852309 on Build GRCh38
rs776646485 Chr.4: 155852149-155852149 on Build GRCh38
rs919484363 Chr.4: 155852176-155852176 on Build GRCh38
rs1000415258 Chr.4: 155852273-155852273 on Build GRCh38
rs528420062 Chr.4: 155852235-155852235 on Build GRCh38
rs201366092 Chr.4: 155852326-155852326 on Build GRCh38
rs772274979 Chr.4: 155852292-155852292 on Build GRCh38
rs755610273 Chr.4: 155852344-155852344 on Build GRCh38
rs746156985 Chr.4: 155852305-155852306 on Build GRCh38
rs779315306 Chr.4: 155852358-155852358 on Build GRCh38
rs151022320 Chr.4: 155852153-155852153 on Build GRCh38
rs750230770 Chr.4: 155852199-155852199 on Build GRCh38
rs772125746 Chr.4: 155852336-155852337 on Build GRCh38
rs751203128 Chr.4: 155852220-155852220 on Build GRCh38
rs371189803 Chr.4: 155852249-155852249 on Build GRCh38
rs145959196 Chr.4: 155852294-155852294 on Build GRCh38
rs760718290 Chr.4: 155852304-155852304 on Build GRCh38
rs367716260 Chr.4: 155852345-155852345 on Build GRCh38
rs570215493 Chr.4: 155852352-155852352 on Build GRCh38
rs759633831 Chr.4: 155852180-155852180 on Build GRCh38
rs761802677 Chr.4: 155852193-155852193 on Build GRCh38
rs746216239 Chr.4: 155852364-155852364 on Build GRCh38
rs776552469 Chr.4: 155852191-155852191 on Build GRCh38
rs752296748 Chr.4: 155852342-155852342 on Build GRCh38
rs373354327 Chr.4: 155852160-155852160 on Build GRCh38
rs765973365 Chr.4: 155852218-155852218 on Build GRCh38
rs376616677 Chr.4: 155852214-155852214 on Build GRCh38
rs376411748 Chr.4: 155852321-155852321 on Build GRCh38
rs775472604 Chr.4: 155852337-155852337 on Build GRCh38
rs774592709 Chr.4: 155852172-155852172 on Build GRCh38
rs142376299 Chr.4: 155852284-155852284 on Build GRCh38
rs758176544 Chr.4: 155852201-155852201 on Build GRCh38
rs781088173 Chr.4: 155852170-155852170 on Build GRCh38
rs774491847 Chr.4: 155852302-155852303 on Build GRCh38
rs767257897 Chr.4: 155852340-155852340 on Build GRCh38
rs369093505 Chr.4: 155852159-155852159 on Build GRCh38
rs759321699 Chr.4: 155852337-155852337 on Build GRCh38
rs767526923 Chr.4: 155852185-155852185 on Build GRCh38
rs373229990 Chr.4: 155852251-155852251 on Build GRCh38
rs776550730 Chr.4: 155852253-155852253 on Build GRCh38
rs747650157 Chr.4: 155852239-155852239 on Build GRCh38

View Full Product Details