Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92088565-92088824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD8
Entrez Gene ID
284697
Gene Name
BTB domain containing 8
Gene Aliases
-
UniGene
Hs.676102
Gene Chromosome Location
Chr.1: 92080305-92147840 on Build GRCh38
Transcripts
NM_183242.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4758819 c.197G>C p.C66S Chr.1: 92088745-92088745 on Build GRCh38
7182593 c.200C>T p.T67I Chr.1: 92088748-92088748 on Build GRCh38
7182594 c.200C>T p.T67I Chr.1: 92088748-92088748 on Build GRCh38
4804903 c.251A>G p.Y84C Chr.1: 92088799-92088799 on Build GRCh38
4804904 c.251A>G p.Y84C Chr.1: 92088799-92088799 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776244655 Chr.1: 92088731-92088731 on Build GRCh38
rs775390943 Chr.1: 92088747-92088747 on Build GRCh38
rs755723349 Chr.1: 92088653-92088653 on Build GRCh38
rs764165055 Chr.1: 92088764-92088764 on Build GRCh38
rs988050636 Chr.1: 92088631-92088631 on Build GRCh38
rs767959237 Chr.1: 92088769-92088769 on Build GRCh38
rs771841607 Chr.1: 92088707-92088707 on Build GRCh38
rs1021882772 Chr.1: 92088642-92088642 on Build GRCh38
rs928456937 Chr.1: 92088613-92088613 on Build GRCh38
rs551733877 Chr.1: 92088694-92088694 on Build GRCh38
rs779405881 Chr.1: 92088656-92088656 on Build GRCh38
rs748995632 Chr.1: 92088674-92088674 on Build GRCh38
rs750957039 Chr.1: 92088649-92088649 on Build GRCh38
rs534518449 Chr.1: 92088768-92088768 on Build GRCh38
rs112190100 Chr.1: 92088679-92088679 on Build GRCh38
rs34856868 Chr.1: 92088726-92088726 on Build GRCh38
rs141744738 Chr.1: 92088724-92088724 on Build GRCh38
rs769966392 Chr.1: 92088742-92088742 on Build GRCh38
rs374624737 Chr.1: 92088752-92088752 on Build GRCh38
rs778778188 Chr.1: 92088685-92088685 on Build GRCh38
rs938562610 Chr.1: 92088628-92088628 on Build GRCh38
rs941989266 Chr.1: 92088657-92088657 on Build GRCh38
rs776495478 Chr.1: 92088649-92088649 on Build GRCh38
rs751817115 Chr.1: 92088765-92088765 on Build GRCh38
rs916002186 Chr.1: 92088643-92088643 on Build GRCh38
rs756527280 Chr.1: 92088783-92088783 on Build GRCh38
rs751013574 Chr.1: 92088780-92088780 on Build GRCh38
rs768156619 Chr.1: 92088680-92088680 on Build GRCh38

View Full Product Details